Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other FOXO1 SIRNA Assays ›
          Gene Symbol:
          FOXO1
          Gene Name:
          forkhead box O1
          Gene Aliases:
          FKH1, FKHR, FOXO1A
          Chromosome Location:
          Chr.13: 40555664 - 40666597 on Build GRCh38
          UniGene ID:
          Hs.370666
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s5259

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information
          • Citations (1)

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: FOXO1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_002015.3 NP_002006.2 2 1346
          XM_011535008.2 XP_011533310.1 2 1499
          XM_011535010.2 XP_011533312.1 2 4011
          GenBank mRNA AF032885.1 - 2 1346
          AK310511.1 - 2 438
          BC021981.2 - 2 1274 2959021
          BC070065.1 - 2 1337 30345006
          BT007455.1 - 2 961
          BX648278.1 - 2 549
          U02310.1 - 2 967

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          blood vessel development
          temperature homeostasis
          cellular glucose homeostasis
          transcription, DNA-templated
          protein acetylation
          autophagy
          apoptotic process
          cellular response to DNA damage stimulus
          insulin receptor signaling pathway
          cellular response to starvation
          positive regulation of autophagy
          endocrine pancreas development
          cellular response to insulin stimulus
          negative regulation of stress-activated MAPK cascade
          cellular response to oxidative stress
          regulation of gluconeogenesis by regulation of transcription from RNA polymerase II promoter
          positive regulation of apoptotic process
          negative regulation of apoptotic process
          fat cell differentiation
          negative regulation of fat cell differentiation
          positive regulation of gluconeogenesis
          positive regulation of protein catabolic process
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          enamel mineralization
          cellular response to hydrogen peroxide
          cellular response to cold
          cellular response to hyperoxia
          cellular response to dexamethasone stimulus
          cellular response to nitric oxide
          negative regulation of canonical Wnt signaling pathway
          neuronal stem cell population maintenance
          response to fluoride
          regulation of neural precursor cell proliferation
          regulation of reactive oxygen species metabolic process
          regulation of energy homeostasis
          RNA polymerase II transcription factor activity, sequence-specific DNA binding
          transcription factor activity, transcription factor binding
          transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
          transcription coactivator binding
          chromatin binding
          protein binding
          beta-catenin binding
          ubiquitin protein ligase binding
          sequence-specific DNA binding
          protein phosphatase 2A binding

          Back To Top
           

          Citations (1)

          FOXO1A is a target for HER2-overexpressing breast tumors.
          Authors
          Wu Y, Shang X, Sarkissyan M, Slamon D, Vadgama JV
          Journal
          Cancer Res - Page No 5475 (Thu Jul 01 00:00:00 UTC 2010)
          Volume (Year)
          70 (2010)
          Product Usage
          -

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-l2qgx:80/100.66.79.25:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0