Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ADAR SIRNA Assays ›
          Gene Symbol:
          ADAR
          Gene Name:
          adenosine deaminase, RNA specific
          Gene Aliases:
          ADAR1, AGS6, DRADA, DSH, DSRAD, G1P1, IFI-4, IFI4, K88DSRBP, P136
          Chromosome Location:
          Chr.1: 154582057 - 154628016 on Build GRCh38
          UniGene ID:
          Hs.12341
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s526070

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ADAR
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001025107.2 NP_001020278.1 2 820
          NM_001111.4 NP_001102.2 2 951
          NM_001193495.1 NP_001180424.1 2 989
          NM_015840.3 NP_056655.2 2 951
          NM_015841.3 NP_056656.2 2 951
          XM_006711109.2 XP_006711172.1 2 757
          XM_006711111.3 XP_006711174.1 3 879
          XM_006711112.2 XP_006711175.1 3 581
          XM_006711113.2 XP_006711176.1 3 655
          XM_011509060.1 XP_011507362.1 2 853
          XM_011509061.2 XP_011507363.2 2 820
          XM_011509062.1 XP_011507364.1 2 780
          XM_017000037.1 XP_016855526.1 3 748
          GenBank mRNA AB209891.1 - 2 764
          AK097241.1 - 3 591
          AK304153.1 - 2 905
          BC017853.1 - 1 284
          BC038227.1 - 2 878 5497548
          BX538232.1 - 2 804
          BX640741.1 - 2 802
          KJ544856.1 - 1 709
          U10439.1 - 2 863
          U18121.1 - 2 756
          X79448.1 - 2 989
          X79449.1 - 2 911
          X98559.1 - 1 694

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          RNA metabolism protein nucleic acid metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          osteoblast differentiation
          in utero embryonic development
          hematopoietic progenitor cell differentiation
          somatic diversification of immune receptors via somatic mutation
          adenosine to inosine editing
          mRNA processing
          response to virus
          base conversion or substitution editing
          erythrocyte differentiation
          pre-miRNA processing
          miRNA loading onto RISC involved in gene silencing by miRNA
          response to interferon-alpha
          negative regulation of apoptotic process
          negative regulation of protein kinase activity by regulation of protein phosphorylation
          positive regulation of viral genome replication
          negative regulation of viral genome replication
          innate immune response
          defense response to virus
          definitive hemopoiesis
          type I interferon signaling pathway
          negative regulation of type I interferon-mediated signaling pathway
          hematopoietic stem cell homeostasis
          cellular response to virus
          negative regulation of RNA interference
          DNA binding
          double-stranded RNA adenosine deaminase activity
          protein binding
          poly(A) RNA binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-r6bjv:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0