Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other FLNA SIRNA Assays ›
          Gene Symbol:
          FLNA
          Gene Name:
          filamin A
          Gene Aliases:
          ABP-280, ABPX, CSBS, CVD1, FLN, FLN-A, FLN1, FMD, MNS, NHBP, OPD, OPD1, OPD2, XLVD, XMVD
          Chromosome Location:
          Chr.X: 154348529 - 154374638 on Build GRCh38
          UniGene ID:
          Hs.195464
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s5276
          Size
          Availability Made To Order
          Catalog #
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: FLNA
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001110556.1 NP_001104026.1 24 4366
          NM_001456.3 NP_001447.2 24 4366
          XM_011531127.1 XP_011529429.1 24 4361
          XM_011531128.1 XP_011529430.1 24 4361
          XM_011531129.1 XP_011529431.1 24 4361
          XM_011531130.1 XP_011529432.1 24 4361
          XM_011531131.1 XP_011529433.1 23 4160
          GenBank mRNA AB191259.1 - 23 4089
          AB191260.1 - 23 4094
          AB371574.1 - 23 4094
          AB371575.1 - 23 4089
          AB371576.1 - 23 4094
          AB371577.1 - 23 4089
          AB371578.1 - 23 4089
          AB371579.1 - 23 4089
          AB593010.1 - 23 4229
          AK074048.1 - 3 566
          AK090427.2 - 23 4155
          AL157419.1 - 1 130
          GU727643.1 - 24 4366
          X53416.1 - 24 4289

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          platelet degranulation
          adenylate cyclase-inhibiting dopamine receptor signaling pathway
          negative regulation of transcription from RNA polymerase I promoter
          platelet activation
          regulation of cell migration
          actin cytoskeleton reorganization
          cell junction assembly
          protein localization to cell surface
          negative regulation of protein catabolic process
          cilium assembly
          mRNA transcription from RNA polymerase II promoter
          positive regulation of transcription factor import into nucleus
          negative regulation of apoptotic process
          receptor clustering
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          negative regulation of sequence-specific DNA binding transcription factor activity
          wound healing, spreading of cells
          establishment of protein localization
          protein stabilization
          cytoplasmic sequestering of protein
          actin crosslink formation
          platelet aggregation
          semaphorin-plexin signaling pathway
          mitotic spindle assembly
          positive regulation of substrate adhesion-dependent cell spreading
          positive regulation of integrin-mediated signaling pathway
          G-protein coupled receptor binding
          glycoprotein binding
          signal transducer activity
          protein binding
          transcription factor binding
          Rho GTPase binding
          Ral GTPase binding
          kinase binding
          small GTPase binding
          mu-type opioid receptor binding
          Fc-gamma receptor I complex binding
          protein homodimerization activity
          poly(A) RNA binding
          SMAD binding
          Rac GTPase binding
          actin filament binding
          GTPase binding
          cadherin binding involved in cell-cell adhesion

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-kkgv7:80/100.66.73.110:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0