Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other WRN SIRNA Assays ›
          Gene Symbol:
          WRN
          Gene Name:
          Werner syndrome RecQ like helicase
          Gene Aliases:
          RECQ3, RECQL2, RECQL3
          Chromosome Location:
          Chr.8: 31033262 - 31173761 on Build GRCh38
          UniGene ID:
          Hs.632050
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s528763

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: WRN
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000553.4 NP_000544.2 2 853
          XM_011544639.2 XP_011542941.1 2 335
          GenBank mRNA AF091214.1 - 2 296
          AY818673.1 - 1 65
          BP279291.1 - 2 246

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          DNA helicase

          Gene Ontology Categories:

          Function(s) Process(es)

          telomere maintenance via recombination
          telomere maintenance
          DNA synthesis involved in DNA repair
          strand displacement
          replicative cell aging
          DNA metabolic process
          DNA replication
          base-excision repair
          double-strand break repair
          cellular response to DNA damage stimulus
          response to oxidative stress
          brain development
          aging
          cell aging
          cellular response to starvation
          response to UV-C
          multicellular organism aging
          protein sumoylation
          replication fork processing
          nucleolus to nucleoplasm transport
          DNA duplex unwinding
          regulation of growth rate
          regulation of apoptotic process
          positive regulation of hydrolase activity
          cellular response to gamma radiation
          nucleic acid phosphodiester bond hydrolysis
          regulation of signal transduction by p53 class mediator
          magnesium ion binding
          Y-form DNA binding
          bubble DNA binding
          DNA binding
          DNA helicase activity
          chromatin binding
          ATP-dependent DNA helicase activity
          helicase activity
          exonuclease activity
          protein binding
          ATP binding
          3'-5' exonuclease activity
          four-way junction helicase activity
          ATPase activity
          manganese ion binding
          protein complex binding
          protein homodimerization activity
          3'-5' DNA helicase activity
          ATP-dependent 3'-5' DNA helicase activity
          G-quadruplex DNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-c56l5:80/100.66.76.41:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0