Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • Enterprise Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other BBS4 SIRNA Assays ›
          Gene Symbol:
          BBS4
          Gene Name:
          Bardet-Biedl syndrome 4
          Gene Aliases:
          -
          Chromosome Location:
          Chr.15: 72686179 - 72738476 on Build GRCh38
          UniGene ID:
          Hs.208681
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s529141

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: BBS4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001252678.1 NP_001239607.1 15 1469
          NM_001320665.1 NP_001307594.1 15 1447
          NM_033028.4 NP_149017.2 16 1516
          NR_045565.1 17 1574
          NR_045566.1 16 1829
          XM_011521848.1 XP_011520150.1 16 1491
          XM_011521849.1 XP_011520151.1 15 1379
          XM_011521851.1 XP_011520153.1 14 1367
          XM_017022450.1 XP_016877939.1 15 1728
          XM_017022451.1 XP_016877940.1 15 1457
          XM_017022452.1 XP_016877941.1 15 1687
          XM_017022453.1 XP_016877942.1 15 1374
          XM_017022454.1 XP_016877943.1 14 1324
          GenBank mRNA AF359281.1 - 16 1488
          AK075321.1 - 16 1480
          AK298922.1 - 17 1546
          AK302224.1 - 16 1783
          AK303706.1 - 15 1464
          AK314507.1 - 16 1488
          AY457143.1 - 16 1588
          BC008923.2 - 13 1293
          BC027624.2 - 16 1486 4824276
          BX647855.1 - 15 1440
          KU177936.1 - 16 1467
          KU177937.1 - 7 780

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          microtubule cytoskeleton organization
          mitotic cytokinesis
          neuron migration
          neural tube closure
          retina homeostasis
          heart looping
          negative regulation of systemic arterial blood pressure
          spermatid development
          visual perception
          sensory perception of smell
          negative regulation of gene expression
          protein transport
          dendrite development
          regulation of lipid metabolic process
          ventricular system development
          striatum development
          hippocampus development
          cerebral cortex development
          adult behavior
          negative regulation of actin filament polymerization
          melanosome transport
          regulation of cytokinesis
          protein localization to organelle
          negative regulation of GTPase activity
          microtubule anchoring at centrosome
          nonmotile primary cilium assembly
          social behavior
          photoreceptor cell outer segment organization
          negative regulation of appetite by leptin-mediated signaling pathway
          positive regulation of multicellular organism growth
          cilium assembly
          fat cell differentiation
          photoreceptor cell maintenance
          positive regulation of cilium assembly
          retinal rod cell development
          intracellular transport
          brain morphogenesis
          sensory processing
          centrosome organization
          maintenance of protein location in nucleus
          regulation of stress fiber assembly
          cilium morphogenesis
          regulation of cilium beat frequency involved in ciliary motility
          face development
          fat pad development
          protein localization to cilium
          protein localization to centrosome
          regulation of nonmotile primary cilium assembly
          protein localization to photoreceptor outer segment
          RNA polymerase II repressing transcription factor binding
          microtubule motor activity
          protein binding
          dynactin binding
          alpha-tubulin binding
          beta-tubulin binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0