Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other BTD SIRNA Assays ›
          Gene Symbol:
          BTD
          Gene Name:
          biotinidase
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 15601352 - 15653709 on Build GRCh38
          UniGene ID:
          Hs.444197
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s529144

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: BTD
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001281726.1 NP_001268655.1 3 432
          XM_006713314.3 XP_006713377.1 4 712
          XM_011534041.2 XP_011532343.1 4 549
          XM_017007088.1 XP_016862577.1 5 825
          GenBank mRNA AK294301.1 - 5 564
          AK297033.1 - 3 424
          AK301838.1 - 3 624
          AK313252.1 - 3 415
          BC012099.1 - 3 358 4559618
          BM839350.1 - 5 515
          BM919775.1 - 3 425
          BP326502.1 - 3 432
          U03274.1 - 3 361
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA DC404964.1 - 3 377 Chr. 3 - 15601801 - 15644362 [+] on Build GRCh38
          GenBankmRNA DC395544.1 - 3 377 Chr. 3 - 15601801 - 15644374 [+] on Build GRCh38
          GenBankmRNA BP289331.1 - 3 429 Chr. 3 - 15601748 - 15644335 [+] on Build GRCh38
          GenBankmRNA DC315657.1 - 3 429 Chr. 3 - 15601748 - 15642055 [+] on Build GRCh38
          GenBankmRNA DC339839.1 - 3 431 Chr. 3 - 15601746 - 15642048 [+] on Build GRCh38
          GenBankmRNA DA670198.1 - 3 308 Chr. 3 - 15601745 - 15644433 [+] on Build GRCh38
          GenBankmRNA DC391100.1 - 3 376 Chr. 3 - 15601801 - 15644346 [+] on Build GRCh38
          GenBankmRNA BP239399.1 - 3 427 Chr. 3 - 15601750 - 15642059 [+] on Build GRCh38
          GenBankmRNA DC383371.1 - 3 419 Chr. 3 - 15601758 - 15642048 [+] on Build GRCh38
          GenBankmRNA HY101778.1 - 3 429 Chr. 3 - 15601749 - 15641949 [+] on Build GRCh38
          GenBankmRNA DA974282.1 - 3 401 Chr. 3 - 15623781 - 15644348 [+] on Build GRCh38
          GenBankmRNA DC379869.1 - 3 430 Chr. 3 - 15601747 - 15642055 [+] on Build GRCh38
          GenBankmRNA DC405860.1 - 3 417 Chr. 3 - 15601760 - 15644348 [+] on Build GRCh38
          GenBankmRNA BQ707863.1 - 2 296 Chr. 3 - 15635421 - 15644817 [+] on Build GRCh38
          GenBankmRNA DC379241.1 - 3 417 Chr. 3 - 15601760 - 15644336 [+] on Build GRCh38
          GenBankmRNA DC386874.1 - 3 419 Chr. 3 - 15601758 - 15642062 [+] on Build GRCh38
          GenBankmRNA DC356757.1 - 3 417 Chr. 3 - 15601760 - 15644339 [+] on Build GRCh38
          GenBankmRNA DC406928.1 - 3 432 Chr. 3 - 15601745 - 15642018 [+] on Build GRCh38
          GenBankmRNA BI834207.1 - 3 411 Chr. 3 - 15601766 - 15644646 [+] on Build GRCh38
          GenBankmRNA DC379106.1 - 3 415 Chr. 3 - 15601762 - 15644338 [+] on Build GRCh38
          GenBankmRNA CB111882.1 - 3 374 Chr. 3 - 15601803 - 15642080 [+] on Build GRCh38
          GenBankmRNA DC380393.1 - 3 420 Chr. 3 - 15601758 - 15642064 [+] on Build GRCh38
          GenBankmRNA DC385352.1 - 3 390 Chr. 3 - 15601788 - 15644354 [+] on Build GRCh38
          GenBankmRNA DC301952.1 - 3 376 Chr. 3 - 15601801 - 15644399 [+] on Build GRCh38
          GenBankmRNA DC388045.1 - 3 377 Chr. 3 - 15601801 - 15644372 [+] on Build GRCh38
          GenBankmRNA HY066319.1 - 3 433 Chr. 3 - 15601745 - 15641967 [+] on Build GRCh38
          GenBankmRNA DA713597.1 - 3 304 Chr. 3 - 15601748 - 15644419 [+] on Build GRCh38
          GenBankmRNA DC346419.1 - 3 429 Chr. 3 - 15601748 - 15642064 [+] on Build GRCh38
          GenBankmRNA BP294125.1 - 3 376 Chr. 3 - 15601801 - 15644388 [+] on Build GRCh38
          GenBankmRNA BP363684.1 - 3 376 Chr. 3 - 15601801 - 15644389 [+] on Build GRCh38
          GenBankmRNA DB179488.1 - 3 306 Chr. 3 - 15601747 - 15644411 [+] on Build GRCh38
          GenBankmRNA DC295436.1 - 3 432 Chr. 3 - 15601745 - 15642064 [+] on Build GRCh38
          GenBankmRNA BP271460.1 - 3 376 Chr. 3 - 15601801 - 15644389 [+] on Build GRCh38
          GenBankmRNA HY179171.1 - 3 312 Chr. 3 - 15601742 - 15644354 [+] on Build GRCh38
          GenBankmRNA DA012679.1 - 3 308 Chr. 3 - 15601745 - 15644444 [+] on Build GRCh38
          GenBankmRNA DC345244.1 - 3 414 Chr. 3 - 15601764 - 15644340 [+] on Build GRCh38
          GenBankmRNA BP212549.1 - 3 417 Chr. 3 - 15601760 - 15644344 [+] on Build GRCh38
          GenBankmRNA DC299579.1 - 3 429 Chr. 3 - 15601748 - 15642059 [+] on Build GRCh38
          GenBankmRNA DC362714.1 - 3 377 Chr. 3 - 15601801 - 15642005 [+] on Build GRCh38
          GenBankmRNA BI761569.1 - 3 360 Chr. 3 - 15601817 - 15644394 [+] on Build GRCh38
          GenBankmRNA BP277224.1 - 3 376 Chr. 3 - 15601802 - 15644367 [+] on Build GRCh38
          GenBankmRNA DC419792.1 - 3 417 Chr. 3 - 15601760 - 15641968 [+] on Build GRCh38
          GenBankmRNA DA621182.1 - 3 306 Chr. 3 - 15601747 - 15644436 [+] on Build GRCh38
          GenBankmRNA DC412448.1 - 3 430 Chr. 3 - 15601747 - 15642038 [+] on Build GRCh38
          GenBankmRNA DA070244.1 - 3 314 Chr. 3 - 15601739 - 15644449 [+] on Build GRCh38
          GenBankmRNA DC397964.1 - 3 376 Chr. 3 - 15601801 - 15644350 [+] on Build GRCh38
          GenBankmRNA AL523081.3 - 2 283 Chr. 3 - 15635423 - 15644857 [+] on Build GRCh38
          GenBankmRNA DC312958.1 - 3 377 Chr. 3 - 15601801 - 15644372 [+] on Build GRCh38
          GenBankmRNA DC399776.1 - 3 377 Chr. 3 - 15601801 - 15642054 [+] on Build GRCh38
          GenBankmRNA BE871868.1 - 2 284 Chr. 3 - 15635422 - 15644646 [+] on Build GRCh38
          GenBankmRNA DC316630.1 - 3 376 Chr. 3 - 15601801 - 15644373 [+] on Build GRCh38
          GenBankmRNA BG323330.1 - 3 359 Chr. 3 - 15601819 - 15644593 [+] on Build GRCh38
          GenBankmRNA DB183961.1 - 3 304 Chr. 3 - 15601749 - 15644455 [+] on Build GRCh38
          GenBankmRNA DC316413.1 - 3 427 Chr. 3 - 15601750 - 15642048 [+] on Build GRCh38
          GenBankmRNA BP344472.1 - 3 304 Chr. 3 - 15601749 - 15644462 [+] on Build GRCh38
          GenBankmRNA HY025659.1 - 3 309 Chr. 3 - 15601745 - 15644358 [+] on Build GRCh38
          GenBankmRNA DC361621.1 - 3 377 Chr. 3 - 15601801 - 15644361 [+] on Build GRCh38
          GenBankmRNA HY122595.1 - 3 431 Chr. 3 - 15601746 - 15641992 [+] on Build GRCh38
          GenBankmRNA BJ993549.1 - 3 376 Chr. 3 - 15601801 - 15644382 [+] on Build GRCh38
          GenBankmRNA DC328386.1 - 3 417 Chr. 3 - 15601760 - 15644338 [+] on Build GRCh38
          GenBankmRNA BM840185.1 - 3 429 Chr. 3 - 15601748 - 15642218 [+] on Build GRCh38
          GenBankmRNA DC393144.1 - 3 377 Chr. 3 - 15601801 - 15644356 [+] on Build GRCh38
          GenBankmRNA DB483481.1 - 3 376 Chr. 3 - 15601801 - 15641998 [+] on Build GRCh38
          GenBankmRNA DC363303.1 - 3 377 Chr. 3 - 15601801 - 15644342 [+] on Build GRCh38
          GenBankmRNA DC327090.1 - 3 429 Chr. 3 - 15601748 - 15642056 [+] on Build GRCh38
          GenBankmRNA DC420042.1 - 3 418 Chr. 3 - 15601760 - 15642000 [+] on Build GRCh38
          GenBankmRNA DC392145.1 - 3 411 Chr. 3 - 15601766 - 15644551 [+] on Build GRCh38
          GenBankmRNA DC358769.1 - 3 417 Chr. 3 - 15601764 - 15644335 [+] on Build GRCh38
          GenBankmRNA DC414836.1 - 3 417 Chr. 3 - 15601760 - 15642056 [+] on Build GRCh38
          GenBankmRNA HY138445.1 - 3 377 Chr. 3 - 15601801 - 15641988 [+] on Build GRCh38
          GenBankmRNA BG769726.1 - 3 357 Chr. 3 - 15601820 - 15642242 [+] on Build GRCh38
          GenBankmRNA DC366546.1 - 3 434 Chr. 3 - 15601619 - 15642064 [+] on Build GRCh38
          GenBankmRNA CB992557.1 - 3 335 Chr. 3 - 15601745 - 15644609 [+] on Build GRCh38
          GenBankmRNA DC382808.1 - 3 432 Chr. 3 - 15601745 - 15642048 [+] on Build GRCh38
          GenBankmRNA BX402565.2 - 3 381 Chr. 3 - 15601796 - 15644714 [+] on Build GRCh38
          GenBankmRNA BP214206.1 - 3 417 Chr. 3 - 15601760 - 15642101 [+] on Build GRCh38
          GenBankmRNA DC359637.1 - 3 419 Chr. 3 - 15601758 - 15642064 [+] on Build GRCh38
          GenBankmRNA CD358893.1 - 3 376 Chr. 3 - 15602217 - 15644643 [+] on Build GRCh38
          GenBankmRNA DA982276.1 - 3 310 Chr. 3 - 15601745 - 15644538 [+] on Build GRCh38
          GenBankmRNA AL549312.3 - 3 371 Chr. 3 - 15601807 - 15644691 [+] on Build GRCh38
          GenBankmRNA BP250812.1 - 3 308 Chr. 3 - 15601745 - 15644456 [+] on Build GRCh38
          GenBankmRNA DC297042.1 - 3 424 Chr. 3 - 15601631 - 15642064 [+] on Build GRCh38
          GenBankmRNA DC362486.1 - 3 417 Chr. 3 - 15601760 - 15641997 [+] on Build GRCh38
          GenBankmRNA DB023397.1 - 3 305 Chr. 3 - 15601748 - 15644435 [+] on Build GRCh38
          GenBankmRNA BP355742.1 - 3 429 Chr. 3 - 15601748 - 15642078 [+] on Build GRCh38
          GenBankmRNA DC393195.1 - 3 343 Chr. 3 - 15601710 - 15644405 [+] on Build GRCh38
          GenBankmRNA DC344914.1 - 3 417 Chr. 3 - 15601760 - 15642041 [+] on Build GRCh38
          GenBankmRNA HY109254.1 - 3 377 Chr. 3 - 15601801 - 15644331 [+] on Build GRCh38
          GenBankmRNA DC421651.1 - 3 429 Chr. 3 - 15601748 - 15642062 [+] on Build GRCh38
          GenBankmRNA DA207338.1 - 3 307 Chr. 3 - 15601748 - 15644443 [+] on Build GRCh38
          GenBankmRNA AU279476.1 - 3 377 Chr. 3 - 15601802 - 15644369 [+] on Build GRCh38
          GenBankmRNA DC327266.1 - 3 377 Chr. 3 - 15601801 - 15644363 [+] on Build GRCh38
          GenBankmRNA DC359535.1 - 3 334 Chr. 3 - 15601721 - 15644346 [+] on Build GRCh38
          GenBankmRNA DC340630.1 - 3 431 Chr. 3 - 15601746 - 15642053 [+] on Build GRCh38
          GenBankmRNA DC329134.1 - 3 430 Chr. 3 - 15601748 - 15642050 [+] on Build GRCh38
          GenBankmRNA DC413286.1 - 3 377 Chr. 3 - 15601801 - 15644387 [+] on Build GRCh38
          GenBankmRNA DC406777.1 - 3 396 Chr. 3 - 15601781 - 15642054 [+] on Build GRCh38
          GenBankmRNA BP213400.1 - 3 427 Chr. 3 - 15601750 - 15642071 [+] on Build GRCh38
          GenBankmRNA DA018451.1 - 3 308 Chr. 3 - 15601745 - 15644423 [+] on Build GRCh38
          GenBankmRNA BG252311.1 - 287 Chr. - - [-] on Build
          GenBankmRNA DA962675.1 - 3 306 Chr. 3 - 15601748 - 15644385 [+] on Build GRCh38
          GenBankmRNA HY151748.1 - 3 418 Chr. 3 - 15601760 - 15641974 [+] on Build GRCh38
          GenBankmRNA DC361738.1 - 3 413 Chr. 3 - 15601764 - 15642060 [+] on Build GRCh38
          GenBankmRNA DT215549.1 - 3 412 Chr. 3 - 15601765 - 15644457 [+] on Build GRCh38
          GenBankmRNA DB006171.1 - 3 308 Chr. 3 - 15601745 - 15644398 [+] on Build GRCh38
          GenBankmRNA BG747472.1 - 3 397 Chr. 3 - 15601778 - 15642023 [+] on Build GRCh38
          GenBankmRNA CV028566.1 - 3 326 Chr. 3 - 15601851 - 15644513 [+] on Build GRCh38
          GenBankmRNA HY048224.1 - 3 326 Chr. 3 - 15601727 - 15644351 [+] on Build GRCh38
          GenBankmRNA BX382242.2 - 3 370 Chr. 3 - 15601807 - 15644734 [+] on Build GRCh38
          GenBankmRNA DC427836.1 - 3 419 Chr. 3 - 15601758 - 15644338 [+] on Build GRCh38
          GenBankmRNA DC386830.1 - 3 429 Chr. 3 - 15601748 - 15642048 [+] on Build GRCh38
          GenBankmRNA DA762289.1 - 3 308 Chr. 3 - 15601745 - 15644451 [+] on Build GRCh38
          GenBankmRNA AL549778.3 - 3 381 Chr. 3 - 15601796 - 15644748 [+] on Build GRCh38
          GenBankmRNA BP216508.1 - 3 308 Chr. 3 - 15601745 - 15642198 [+] on Build GRCh38
          GenBankmRNA BP321544.1 - 3 305 Chr. 3 - 15601748 - 15644457 [+] on Build GRCh38
          GenBankmRNA DC421473.1 - 3 408 Chr. 3 - 15601769 - 15642062 [+] on Build GRCh38
          GenBankmRNA DC419424.1 - 3 409 Chr. 3 - 15601769 - 15644590 [+] on Build GRCh38
          GenBankmRNA BI560515.1 - 3 594 Chr. 3 - 15602003 - 15644392 [+] on Build GRCh38
          GenBankmRNA BX487803.1 - 3 401 Chr. 3 - 15601777 - 15644340 [+] on Build GRCh38
          GenBankmRNA DA626404.1 - 3 378 Chr. 3 - 15601801 - 15644442 [+] on Build GRCh38
          GenBankmRNA DC366654.1 - 3 377 Chr. 3 - 15601801 - 15644381 [+] on Build GRCh38
          GenBankmRNA DC358370.1 - 3 429 Chr. 3 - 15601748 - 15642045 [+] on Build GRCh38
          GenBankmRNA BP224073.1 - 3 418 Chr. 3 - 15601759 - 15641984 [+] on Build GRCh38
          GenBankmRNA BP332044.1 - 3 308 Chr. 3 - 15601745 - 15644444 [+] on Build GRCh38
          GenBankmRNA HY145750.1 - 3 306 Chr. 3 - 15601747 - 15644340 [+] on Build GRCh38
          GenBankmRNA BP229719.1 - 3 417 Chr. 3 - 15601761 - 15644347 [+] on Build GRCh38
          GenBankmRNA DC326916.1 - 3 413 Chr. 3 - 15601764 - 15644338 [+] on Build GRCh38
          GenBankmRNA DA149430.1 - 3 311 Chr. 3 - 15601745 - 15644448 [+] on Build GRCh38
          GenBankmRNA DC425366.1 - 3 384 Chr. 3 - 15601793 - 15644361 [+] on Build GRCh38
          GenBankmRNA DB261455.1 - 3 305 Chr. 3 - 15601748 - 15644432 [+] on Build GRCh38
          GenBankmRNA DA279665.1 - 3 310 Chr. 3 - 15601745 - 15644362 [+] on Build GRCh38
          GenBankmRNA BI838922.1 - 3 381 Chr. 3 - 15601803 - 15644432 [+] on Build GRCh38
          GenBankmRNA HY134550.1 - 3 431 Chr. 3 - 15601746 - 15641967 [+] on Build GRCh38
          GenBankmRNA BM473862.1 - 3 404 Chr. 3 - 15601774 - 15644777 [+] on Build GRCh38
          GenBankmRNA DA379620.1 - 3 305 Chr. 3 - 15601748 - 15644417 [+] on Build GRCh38
          GenBankmRNA DC388270.1 - 3 430 Chr. 3 - 15601747 - 15642049 [+] on Build GRCh38
          GenBankmRNA DC429240.1 - 3 434 Chr. 3 - 15601745 - 15642062 [+] on Build GRCh38
          GenBankmRNA DC379486.1 - 3 419 Chr. 3 - 15601761 - 15642058 [+] on Build GRCh38
          GenBankmRNA DC405500.1 - 3 415 Chr. 3 - 15601762 - 15644346 [+] on Build GRCh38
          GenBankmRNA AU137728.1 - 3 375 Chr. 3 - 15601803 - 15644578 [+] on Build GRCh38
          GenBankmRNA DC413182.1 - 3 419 Chr. 3 - 15601760 - 15644334 [+] on Build GRCh38
          GenBankmRNA DC336876.1 - 3 417 Chr. 3 - 15601764 - 15642056 [+] on Build GRCh38
          GenBankmRNA CN481922.1 - 3 401 Chr. 3 - 15601776 - 15644406 [+] on Build GRCh38
          GenBankmRNA DA838233.1 - 3 309 Chr. 3 - 15601745 - 15644415 [+] on Build GRCh38
          GenBankmRNA DC383345.1 - 3 413 Chr. 3 - 15601764 - 15642023 [+] on Build GRCh38
          GenBankmRNA BP255505.1 - 3 383 Chr. 3 - 15601794 - 15644381 [+] on Build GRCh38
          GenBankmRNA DB001766.1 - 3 310 Chr. 3 - 15601743 - 15644415 [+] on Build GRCh38
          GenBankmRNA DC295693.1 - 3 432 Chr. 3 - 15601746 - 15642062 [+] on Build GRCh38
          GenBankmRNA DA866082.1 - 3 431 Chr. 3 - 15601747 - 15642010 [+] on Build GRCh38
          GenBankmRNA DA928653.1 - 3 307 Chr. 3 - 15601746 - 15644433 [+] on Build GRCh38
          GenBankmRNA CK905568.1 - 3 501 Chr. 3 - 15632649 - 15641985 [+] on Build GRCh38
          GenBankmRNA DC421370.1 - 3 327 Chr. 3 - 15601726 - 15644444 [+] on Build GRCh38
          GenBankmRNA DA466173.1 - 3 309 Chr. 3 - 15601745 - 15644440 [+] on Build GRCh38
          GenBankmRNA DC326889.1 - 3 427 Chr. 3 - 15601750 - 15642054 [+] on Build GRCh38
          GenBankmRNA BX340297.2 - 3 384 Chr. 3 - 15601793 - 15644390 [+] on Build GRCh38
          GenBankmRNA DA262451.1 - 3 306 Chr. 3 - 15601747 - 15644439 [+] on Build GRCh38
          GenBankmRNA BI759206.1 - 3 368 Chr. 3 - 15601808 - 15644649 [+] on Build GRCh38
          GenBankmRNA DC412266.1 - 3 422 Chr. 3 - 15601760 - 15642060 [+] on Build GRCh38
          GenBankmRNA DC326297.1 - 3 432 Chr. 3 - 15601745 - 15642060 [+] on Build GRCh38
          GenBankmRNA BI770329.1 - 3 419 Chr. 3 - 15601757 - 15644559 [+] on Build GRCh38
          GenBankmRNA BM507279.1 - 3 501 Chr. 3 - 15632649 - 15642017 [+] on Build GRCh38
          GenBankmRNA BP274875.1 - 3 376 Chr. 3 - 15601801 - 15644363 [+] on Build GRCh38
          GenBankmRNA DC355003.1 - 3 376 Chr. 3 - 15601801 - 15644368 [+] on Build GRCh38
          GenBankmRNA HY141700.1 - 3 377 Chr. 3 - 15601801 - 15641989 [+] on Build GRCh38
          GenBankmRNA DC420548.1 - 3 429 Chr. 3 - 15601748 - 15642064 [+] on Build GRCh38
          GenBankmRNA DA044837.1 - 3 303 Chr. 3 - 15601750 - 15644436 [+] on Build GRCh38
          GenBankmRNA DC319971.1 - 3 430 Chr. 3 - 15601748 - 15642056 [+] on Build GRCh38
          GenBankmRNA DC398513.1 - 3 378 Chr. 3 - 15601675 - 15644377 [+] on Build GRCh38
          GenBankmRNA DC297301.1 - 3 432 Chr. 3 - 15601745 - 15642060 [+] on Build GRCh38
          GenBankmRNA DC421150.1 - 3 430 Chr. 3 - 15601747 - 15642064 [+] on Build GRCh38
          GenBankmRNA DA535138.1 - 3 310 Chr. 3 - 15601745 - 15644428 [+] on Build GRCh38

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          hydrolase

          Gene Ontology Categories:

          Function(s) Process(es)

          biotin metabolic process
          nitrogen compound metabolic process
          central nervous system development
          hydrolase activity, acting on carbon-nitrogen (but not peptide) bonds
          biotinidase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8kj9j:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0