Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NOS1 SIRNA Assays ›
          Gene Symbol:
          NOS1
          Gene Name:
          nitric oxide synthase 1
          Gene Aliases:
          IHPS1, N-NOS, NC-NOS, NOS, bNOS, nNOS
          Chromosome Location:
          Chr.12: 117208142 - 117441843 on Build GRCh38
          UniGene ID:
          Hs.654410
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s529224

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NOS1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000620.4 NP_000611.1 7 2051
          NM_001204213.1 NP_001191142.1 6 643
          NM_001204214.1 NP_001191143.1 6 638
          NM_001204218.1 NP_001191147.1 7 2051
          XM_011538398.2 XP_011536700.1 8 1922
          XM_017019345.1 XP_016874834.1 7 2058
          XM_017019346.1 XP_016874835.1 7 1872
          XM_017019347.1 XP_016874836.1 7 1961
          GenBank mRNA AK294435.1 - 7 1855
          AK307481.1 - 7 2051
          D16408.1 - 6 1340
          EU753241.1 - 7 2025
          L02881.1 - 6 1767
          U17327.1 - 7 2025

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: X59949, U67309.

          Panther Classification:

          Molecular Function -

          oxidoreductase

          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          regulation of sodium ion transport
          arginine catabolic process
          nitric oxide biosynthetic process
          striated muscle contraction
          nitric oxide mediated signal transduction
          myoblast fusion
          female pregnancy
          aging
          negative regulation of cell proliferation
          response to heat
          response to lead ion
          negative regulation of calcium ion transport into cytosol
          response to activity
          peptidyl-cysteine S-nitrosylation
          positive regulation of guanylate cyclase activity
          response to lipopolysaccharide
          response to vitamin E
          multicellular organismal response to stress
          positive regulation of histone acetylation
          response to nicotine
          neurotransmitter biosynthetic process
          exogenous drug catabolic process
          negative regulation of apoptotic process
          negative regulation of potassium ion transport
          response to peptide hormone
          response to estrogen
          cell redox homeostasis
          response to ethanol
          negative regulation of blood pressure
          negative regulation of heart contraction
          positive regulation of transcription, DNA-templated
          negative regulation of vasoconstriction
          positive regulation of vasodilation
          positive regulation of transcription from RNA polymerase II promoter
          negative regulation of insulin secretion
          behavioral response to cocaine
          regulation of neurogenesis
          negative regulation of hydrolase activity
          negative regulation of cytosolic calcium ion concentration
          negative regulation of serotonin uptake
          negative regulation of calcium ion transport
          regulation of sensory perception of pain
          oxidation-reduction process
          regulation of cardiac muscle contraction
          regulation of ryanodine-sensitive calcium-release channel activity
          cellular response to mechanical stimulus
          cellular response to growth factor stimulus
          response to nitric oxide
          positive regulation of the force of heart contraction
          positive regulation of long-term synaptic potentiation
          negative regulation of adrenergic receptor signaling pathway involved in heart process
          positive regulation of adrenergic receptor signaling pathway involved in heart process
          positive regulation of neuron death
          positive regulation of sodium ion transmembrane transport
          regulation of calcium ion transmembrane transport via high voltage-gated calcium channel
          regulation of cardiac conduction
          nitric-oxide synthase activity
          iron ion binding
          protein binding
          calmodulin binding
          FMN binding
          sodium channel regulator activity
          enzyme binding
          heme binding
          tetrahydrobiopterin binding
          arginine binding
          protein homodimerization activity
          ion channel binding
          cadmium ion binding
          flavin adenine dinucleotide binding
          NADP binding
          scaffold protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-pnd5g:80/100.66.79.83:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline