Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SMN1 SIRNA Assays ›
          Gene Symbol:
          SMN1 SMN2
          Gene Name:
          survival of motor neuron 1, telomeric
          survival of motor neuron 2, centromeric
          Gene Aliases:
          BCD541, GEMIN1, SMA, SMA1, SMA2, SMA3, SMA4, SMA@, SMN, SMNT, T-BCD541, TDRD16A
          BCD541, C-BCD541, GEMIN1, SMNC, TDRD16B
          Chromosome Location:
          Chr.5: 70924941 - 70953015 on Build GRCh38
          Chromosome Location:
          Chr.5: 70049523 - 70077595 on Build GRCh38
          UniGene ID:
          Hs.535788
          Hs.202179
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID s530580

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SMN1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000344.3 NP_000335.1 7 910
          NM_001297715.1 NP_001284644.1 7 910
          NM_022874.2 NP_075012.1 6 814
          XM_011543596.1 XP_011541898.1 7 789
          XM_011543597.1 XP_011541899.1 6 594
          XM_011543598.2 XP_011541900.1 5 509
          XM_017009786.1 XP_016865275.1 6 693
          GenBank mRNA AK298203.1 - 6 594
          BC062723.1 - 7 766 4250429
          JQ657798.1 - 4 403
          JQ657799.1 - 4 403
          JQ657800.1 - 4 298
          JQ657802.1 - 4 403
          JQ657803.1 - 3 202
          JQ690867.1 - 4 298
          JQ690868.1 - 3 202
          JQ745297.1 - 3 145
          U18423.1 - 7 780
          Gene Symbol: SMN2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_017411.3 NP_059107.1 7 910
          NM_022875.2 NP_075013.1 7 910
          NM_022876.2 NP_075014.1 6 814
          NM_022877.2 NP_075015.1 6 814
          XM_011543599.1 XP_011541901.1 7 789
          XM_011543600.2 XP_011541902.1 6 594
          XM_011543601.1 XP_011541903.1 6 594
          XM_011543602.2 XP_011541904.1 5 510
          XM_011543603.2 XP_011541905.1 5 510
          XM_017009787.1 XP_016865276.1 7 789
          GenBank mRNA AA029652.1 - 4 300
          AK289669.1 - 7 793
          BC000908.2 - 7 779 2985046
          BC015308.1 - 7 777 4427000
          BC070242.1 - 7 756 4673318
          CN272420.1 - 5 613
          JQ690861.1 - 4 298
          JQ690862.1 - 4 298
          JQ690863.1 - 3 202
          JQ690864.1 - 4 346
          JQ690865.1 - 4 403
          JQ690866.1 - 3 202
          U21914.1 - 7 744

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          RNA splicing factor

          Gene Ontology Categories:

          Function(s) Process(es)

          spliceosomal complex assembly
          spliceosomal snRNP assembly
          DNA-templated transcription, termination
          nervous system development
          nuclear import
          RNA binding
          protein binding
          identical protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-mfr4m:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0