Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PLA2G6 SIRNA Assays ›
Gene Symbol:
PLA2G6
Gene Name:
phospholipase A2 group VI
Gene Aliases:
CaI-PLA2, GVI, INAD1, IPLA2-VIA, NBIA2, NBIA2A, NBIA2B, PARK14, PLA2, PNPLA9, iPLA2, iPLA2beta
Chromosome Location:
Chr.22: 38111495 - 38192099 on Build GRCh38
UniGene ID:
Hs.170479
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s530880
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PLA2G6
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001004426.1
NP_001004426.1
2
185
NM_001199562.1
NP_001186491.1
2
140
NM_003560.2
NP_003551.2
2
185
XM_005261764.2
XP_005261821.1
2
160
XM_005261765.1
XP_005261822.1
2
157
XM_005261766.1
XP_005261823.1
2
252
XM_006724332.3
XP_006724395.1
2
161
XM_011530422.1
XP_011528724.1
2
141
XM_011530423.1
XP_011528725.1
2
141
XM_011530424.1
XP_011528726.1
2
139
XM_011530426.2
XP_011528728.1
2
160
XM_017028981.1
XP_016884470.1
2
252
XM_017028982.1
XP_016884471.1
2
157
XM_017028983.1
XP_016884472.1
2
139
XM_017028984.1
XP_016884473.1
2
141
XM_017028985.1
XP_016884474.1
2
116
XM_017028986.1
XP_016884475.1
2
160
XM_017028987.1
XP_016884476.1
2
160
XM_017028988.1
XP_016884477.1
2
160
GenBank mRNA
AF064594.1
-
2
185
AF102988.1
-
1
49
AF102989.1
-
1
49
AK096521.1
-
2
135
AK290825.1
-
2
142
AK291212.1
-
2
171
AK300459.1
-
2
141
AK308308.1
-
2
207
AL080187.1
-
2
185
BC036742.1
-
2
179
5166749
BC051904.1
-
2
121
5175768
CR456543.1
-
2
113
Back To Top
More Information
Panther Classification:
Molecular Function -
phospholipase
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of protein phosphorylation
chemotaxis
positive regulation of cytosolic calcium ion concentration
memory
urinary bladder smooth muscle contraction
lipid catabolic process
antibacterial humoral response
cardiolipin biosynthetic process
response to endoplasmic reticulum stress
positive regulation of insulin secretion involved in cellular response to glucose stimulus
phosphatidylcholine acyl-chain remodeling
phosphatidylethanolamine acyl-chain remodeling
Fc-gamma receptor signaling pathway involved in phagocytosis
positive regulation of vasodilation
positive regulation of exocytosis
negative regulation of synaptic transmission, glutamatergic
maternal process involved in female pregnancy
positive regulation of protein kinase C signaling
positive regulation of release of cytochrome c from mitochondria
positive regulation of arachidonic acid secretion
regulation of store-operated calcium channel activity
positive regulation of ceramide biosynthetic process
phospholipase A2 activity
calmodulin binding
hydrolase activity
protein kinase binding
ATP-dependent protein binding
calcium-independent phospholipase A2 activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0