Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • Enterprise Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PARK2 SIRNA Assays ›
          Gene Symbol:
          PARK2
          Gene Name:
          parkin RBR E3 ubiquitin protein ligase
          Gene Aliases:
          AR-JP, LPRS2, PDJ, PRKN
          Chromosome Location:
          Chr.6: 161347417 - 162727802 on Build GRCh38
          UniGene ID:
          Hs.132954
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s530943

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PARK2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_013988.2 NP_054643.2 2, 3 308

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          protein polyubiquitination
          mitochondrial fission
          mitophagy
          negative regulation of protein phosphorylation
          startle response
          transcription, DNA-templated
          protein monoubiquitination
          response to oxidative stress
          mitochondrion organization
          central nervous system development
          learning
          adult locomotory behavior
          proteasomal protein catabolic process
          regulation of autophagy
          positive regulation of gene expression
          negative regulation of gene expression
          positive regulation of mitochondrial fusion
          negative regulation of mitochondrial fusion
          regulation of mitochondrion organization
          regulation of glucose metabolic process
          free ubiquitin chain polymerization
          regulation of dopamine secretion
          macroautophagy
          protein ubiquitination
          regulation of protein ubiquitination
          protein destabilization
          negative regulation of actin filament bundle assembly
          regulation of lipid transport
          positive regulation of proteasomal ubiquitin-dependent protein catabolic process
          negative regulation of glucokinase activity
          cellular response to unfolded protein
          response to endoplasmic reticulum stress
          synaptic transmission, glutamatergic
          protein K29-linked ubiquitination
          ERAD pathway
          regulation of dopamine metabolic process
          norepinephrine metabolic process
          dopamine metabolic process
          protein ubiquitination involved in ubiquitin-dependent protein catabolic process
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          proteasome-mediated ubiquitin-dependent protein catabolic process
          positive regulation of DNA binding
          negative regulation of neuron apoptotic process
          cellular protein catabolic process
          cellular protein metabolic process
          protein K27-linked ubiquitination
          negative regulation by host of viral genome replication
          positive regulation of protein catabolic process
          positive regulation of transcription from RNA polymerase II promoter
          negative regulation of JNK cascade
          negative regulation of insulin secretion
          protein stabilization
          positive regulation of neurotransmitter uptake
          dopamine uptake involved in synaptic transmission
          protein autoubiquitination
          regulation of mitochondrial membrane potential
          zinc ion homeostasis
          negative regulation of cell death
          regulation of canonical Wnt signaling pathway
          parkin-mediated mitophagy in response to mitochondrial depolarization
          neuron cellular homeostasis
          protein K63-linked ubiquitination
          protein localization to mitochondrion
          aggresome assembly
          protein K48-linked ubiquitination
          protein K11-linked ubiquitination
          cellular response to manganese ion
          protein K6-linked ubiquitination
          negative regulation of canonical Wnt signaling pathway
          positive regulation of mitochondrial fission
          negative regulation of release of cytochrome c from mitochondria
          cellular response to toxic substance
          mitophagy in response to mitochondrial depolarization
          regulation of cellular response to oxidative stress
          negative regulation of neuron death
          positive regulation of proteasomal protein catabolic process
          negative regulation of endoplasmic reticulum stress-induced intrinsic apoptotic signaling pathway
          negative regulation of intrinsic apoptotic signaling pathway by p53 class mediator
          negative regulation of primary amine oxidase activity
          positive regulation of protein linear polyubiquitination
          regulation of synaptic vesicle transport
          negative regulation of oxidative stress-induced cell death
          regulation of protein targeting to mitochondrion
          positive regulation of tumor necrosis factor-mediated signaling pathway
          cellular response to dopamine
          negative regulation of oxidative stress-induced neuron intrinsic apoptotic signaling pathway
          positive regulation of oxidative stress-induced neuron intrinsic apoptotic signaling pathway
          negative regulation of endoplasmic reticulum stress-induced neuron intrinsic apoptotic signaling pathway
          positive regulation of dendrite extension
          negative regulation of spontaneous neurotransmitter secretion
          regulation of reactive oxygen species metabolic process
          negative regulation of reactive oxygen species metabolic process
          transcription regulatory region sequence-specific DNA binding
          G-protein coupled receptor binding
          transcription factor activity, sequence-specific DNA binding
          actin binding
          ubiquitin-protein transferase activity
          protein binding
          beta-catenin binding
          zinc ion binding
          tubulin binding
          ligase activity
          SH3 domain binding
          enzyme binding
          kinase binding
          protein kinase binding
          PDZ domain binding
          Hsp70 protein binding
          heat shock protein binding
          ubiquitin conjugating enzyme binding
          ubiquitin protein ligase binding
          identical protein binding
          histone deacetylase binding
          ubiquitin binding
          phospholipase binding
          chaperone binding
          ubiquitin protein ligase activity
          cullin family protein binding
          ubiquitin protein ligase activity involved in ERAD pathway
          ubiquitin-specific protease binding
          F-box domain binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-p9bc6:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0