Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: KAZN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201628.2 | NP_963922.2 | 12, 13 | 2157 | ||
XM_005245795.4 | XP_005245852.1 | 14, 15 | 2412 | |||
XM_011541074.2 | XP_011539376.1 | 14, 15 | 2419 | |||
XM_011541075.2 | XP_011539377.1 | 12, 13 | 2075 | |||
XM_011541076.2 | XP_011539378.1 | 12, 13 | 2220 | |||
XM_011541077.2 | XP_011539379.1 | 12, 13 | 3199 | |||
XM_011541081.2 | XP_011539383.1 | 6, 7 | 899 | |||
GenBank mRNA | AK293806.1 | - | 7, 8 | 963 | ||
BC101637.1 | - | 2, 3 | 238 | |||
BC113621.1 | - | 2, 3 | 238 | |||
BC143641.1 | - | 2, 3 | 184 | |||
DA317442.1 | - | 2, 3 | 175 | |||
DN831635.1 | - | 3, 4 | 439 |