Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Enterprise Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other AXIN2 SIRNA Assays ›
          Gene Symbol:
          AXIN2
          Gene Name:
          axin 2
          Gene Aliases:
          AXIL, ODCRCS
          Chromosome Location:
          Chr.17: 65528563 - 65561622 on Build GRCh38
          UniGene ID:
          Hs.156527
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s531449

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: AXIN2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_004655.3 NP_004646.3 6 1604
          XM_011525319.2 XP_011523621.1 6 1815
          XM_011525320.1 XP_011523622.1 6 1804
          XM_011525321.2 XP_011523623.1 6 1752
          XM_017025192.1 XP_016880681.1 6 1471
          XM_017025193.1 XP_016880682.1 6 1804
          XM_017025194.1 XP_016880683.1 6 1583
          GenBank mRNA AF078165.1 - 5 1404
          AF205888.1 - 5 1421
          AK025718.1 - 5 626
          AK315303.1 - 6 1520
          BC101533.1 - 5 1384 8069039
          BC143244.1 - 5 1384 9051745
          CR933657.1 - 6 1629
          DA839648.1 - 1 82

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of protein phosphorylation
          intramembranous ossification
          secondary heart field specification
          chondrocyte differentiation involved in endochondral bone morphogenesis
          regulation of transcription, DNA-templated
          cell proliferation
          negative regulation of cell proliferation
          dorsal/ventral axis specification
          positive regulation of epithelial to mesenchymal transition
          positive regulation of cell death
          Wnt signaling pathway
          bone mineralization
          regulation of mismatch repair
          cellular protein localization
          negative regulation of catenin import into nucleus
          odontogenesis
          positive regulation of GTPase activity
          maintenance of DNA repeat elements
          negative regulation of osteoblast differentiation
          mRNA stabilization
          regulation of chondrocyte development
          regulation of centromeric sister chromatid cohesion
          cellular response to organic cyclic compound
          negative regulation of canonical Wnt signaling pathway
          Wnt signaling pathway involved in somitogenesis
          positive regulation of canonical Wnt signaling pathway
          beta-catenin-TCF complex assembly
          GTPase activator activity
          protein binding
          beta-catenin binding
          enzyme binding
          protein kinase binding
          ubiquitin protein ligase binding
          armadillo repeat domain binding
          I-SMAD binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0