Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CASP8 SIRNA Assays ›
          Gene Symbol:
          CASP8
          Gene Name:
          caspase 8
          Gene Aliases:
          ALPS2B, CAP4, Casp-8, FLICE, MACH, MCH5
          Chromosome Location:
          Chr.2: 201233443 - 201287711 on Build GRCh38
          UniGene ID:
          Hs.599762
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s531490

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CASP8
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001080124.1 NP_001073593.1 6 833
          NM_001080125.1 NP_001073594.1 6 1021
          NM_001228.4 NP_001219.2 7 997
          NM_033355.3 NP_203519.1 7 852
          NM_033356.3 NP_203520.1 5 738
          NR_111983.1 6 1021
          XM_005246885.1 XP_005246942.1 5 796
          XM_005246886.1 XP_005246943.1 6 767
          XM_005246887.1 XP_005246944.1 7 919
          XM_005246888.1 XP_005246945.1 7 861
          XM_005246889.1 XP_005246946.1 6 738
          XM_005246890.3 XP_005246947.1 6 745
          XM_005246891.4 XP_005246948.1 6 985
          XM_005246892.1 XP_005246949.1 5 693
          XM_006712789.1 XP_006712852.1 7 878
          XM_006712790.3 XP_006712853.1 6 757
          XM_011511969.1 XP_011510271.1 4 322
          GenBank mRNA AB451282.1 - 4 598
          AB451414.1 - 4 598
          AF009620.1 - 4 598
          AF380342.1 - 6 728
          AF422927.1 - 6 821
          AF422928.1 - 7 840
          BC028223.2 - 5 670
          BC068050.1 - 5 688 6421760
          BX395849.2 - 5 687
          U58143.1 - 5 643
          U60520.1 - 7 950
          X98172.1 - 6 934
          X98173.1 - 5 670
          X98177.1 - 5 643
          X98178.1 - 4 598
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA CR975705.1 - 3 157 Chr. 2 - 201272713 - 201285219 [+] on Build GRCh38
          GenBankmRNA BQ949004.1 - 2 108 Chr. 2 - 201272896 - 201285292 [+] on Build GRCh38

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          protease

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          apoptotic process
          activation of cysteine-type endopeptidase activity involved in apoptotic process
          cellular component disassembly involved in execution phase of apoptosis
          cell surface receptor signaling pathway
          response to cold
          regulation of tumor necrosis factor-mediated signaling pathway
          regulation of necrotic cell death
          natural killer cell activation
          macrophage differentiation
          response to cobalt ion
          response to estradiol
          response to lipopolysaccharide
          response to tumor necrosis factor
          TRIF-dependent toll-like receptor signaling pathway
          TRAIL-activated apoptotic signaling pathway
          suppression by virus of host cysteine-type endopeptidase activity involved in apoptotic process
          T cell activation
          B cell activation
          positive regulation of I-kappaB kinase/NF-kappaB signaling
          negative regulation of I-kappaB kinase/NF-kappaB signaling
          response to ethanol
          positive regulation of macrophage differentiation
          positive regulation of proteolysis
          response to antibiotic
          protein heterooligomerization
          proteolysis involved in cellular protein catabolic process
          syncytiotrophoblast cell differentiation involved in labyrinthine layer development
          nucleotide-binding oligomerization domain containing signaling pathway
          cellular response to mechanical stimulus
          cellular response to organic cyclic compound
          death-inducing signaling complex assembly
          apoptotic signaling pathway
          extrinsic apoptotic signaling pathway
          execution phase of apoptosis
          activation of cysteine-type endopeptidase activity
          activation of cysteine-type endopeptidase activity involved in apoptotic signaling pathway
          positive regulation of protein insertion into mitochondrial membrane involved in apoptotic signaling pathway
          regulation of extrinsic apoptotic signaling pathway via death domain receptors
          negative regulation of extrinsic apoptotic signaling pathway via death domain receptors
          cysteine-type endopeptidase activity
          death receptor binding
          tumor necrosis factor receptor binding
          protein binding
          peptidase activity
          cysteine-type peptidase activity
          ubiquitin protein ligase binding
          protein complex binding
          death effector domain binding
          identical protein binding
          scaffold protein binding
          cysteine-type endopeptidase activity involved in apoptotic process
          cysteine-type endopeptidase activity involved in apoptotic signaling pathway

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8xshn:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0