Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other AXIN1 SIRNA Assays ›
          Gene Symbol:
          AXIN1
          Gene Name:
          axin 1
          Gene Aliases:
          AXIN, PPP1R49
          Chromosome Location:
          Chr.16: 287440 - 355226 on Build GRCh38
          UniGene ID:
          Hs.592082
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s531576

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: AXIN1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_003502.3 NP_003493.1 2 676
          NM_181050.2 NP_851393.1 2 676
          NR_134879.1 2 676
          XM_011522682.2 XP_011520984.1 2 687
          XM_011522683.2 XP_011520985.1 2 686
          XM_011522684.2 XP_011520986.1 2 537
          XM_017023743.1 XP_016879232.1 2 687
          XM_017023744.1 XP_016879233.1 2 686
          XM_017023747.1 XP_016879236.1 2 689
          XM_017023748.1 XP_016879237.1 2 689
          GenBank mRNA AF009674.1 - 2 399
          BC044648.1 - 2 448 5809104
          KF615850.1 - 2 676
          KF615851.1 - 2 676
          KF615852.1 - 2 676
          KU178413.1 - 1 287
          KU178414.1 - 1 287
          KU178415.1 - 1 287

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          optic placode formation
          positive regulation of protein phosphorylation
          apoptotic process
          activation of JUN kinase activity
          determination of left/right symmetry
          cell death
          dorsal/ventral axis specification
          positive regulation of peptidyl-threonine phosphorylation
          Wnt signaling pathway
          forebrain anterior/posterior pattern specification
          Wnt-activated signaling pathway involved in forebrain neuron fate commitment
          negative regulation of Wnt signaling pathway
          olfactory placode formation
          cytoplasmic microtubule organization
          positive regulation of protein ubiquitination
          activation of protein kinase activity
          positive regulation of peptidyl-serine phosphorylation
          negative regulation of transcription elongation from RNA polymerase II promoter
          regulation of catenin import into nucleus
          proteasome-mediated ubiquitin-dependent protein catabolic process
          positive regulation of GTPase activity
          cellular protein complex assembly
          negative regulation of fat cell differentiation
          positive regulation of protein catabolic process
          positive regulation of transcription, DNA-templated
          positive regulation of JNK cascade
          embryonic eye morphogenesis
          axial mesoderm formation
          positive regulation of ubiquitin-protein transferase activity
          muscle cell development
          canonical Wnt signaling pathway
          embryonic skeletal joint morphogenesis
          canonical Wnt signaling pathway involved in neural plate anterior/posterior pattern formation
          cellular response to organic cyclic compound
          negative regulation of canonical Wnt signaling pathway
          Wnt signaling pathway involved in somitogenesis
          positive regulation of canonical Wnt signaling pathway
          beta-catenin destruction complex assembly
          beta-catenin destruction complex disassembly
          positive regulation of protein ubiquitination involved in ubiquitin-dependent protein catabolic process
          signal transducer activity
          GTPase activator activity
          protein binding
          beta-catenin binding
          enzyme binding
          protein kinase binding
          receptor signaling complex scaffold activity
          ubiquitin protein ligase binding
          protein complex scaffold
          identical protein binding
          protein homodimerization activity
          SMAD binding
          armadillo repeat domain binding
          I-SMAD binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0