Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PEX5 SIRNA Assays ›
          Gene Symbol:
          PEX5
          Gene Name:
          peroxisomal biogenesis factor 5
          Gene Aliases:
          PBD2A, PBD2B, PTS1-BP, PTS1R, PXR1, RCDP5
          Chromosome Location:
          Chr.12: 7188072 - 7218574 on Build GRCh38
          UniGene ID:
          Hs.567327
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s532232

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 392.00
          Your Price
          Online offer: 380.65
          380.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PEX5
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000319.4 NP_000310.2 5, 6 567
          NM_001131023.1 NP_001124495.1 6, 7 677
          NM_001131024.1 NP_001124496.1 6, 7 632
          NM_001131025.1 NP_001124497.1 6, 7 1098
          NM_001131026.1 NP_001124498.1 7, 8 1131
          NM_001300789.1 NP_001287718.1 6, 7 703
          XM_005253455.1 XP_005253512.1 6, 7 698
          XM_011520793.2 XP_011519095.1 6, 7 692
          XM_011520795.1 XP_011519097.1 6, 7 1109
          XM_011520797.1 XP_011519099.1 6, 7 602
          XM_011520798.1 XP_011519100.1 7, 8 811
          XM_011520799.2 XP_011519101.1 6, 7 1251
          XM_011520800.1 XP_011519102.1 6, 7 697
          XM_011520801.2 XP_011519103.1 6, 7 691
          XM_011520802.2 XP_011519104.1 5, 6 454
          XM_017019744.1 XP_016875233.1 5, 6 1036
          XM_017019745.1 XP_016875234.1 5, 6 1037
          XM_017019746.1 XP_016875235.1 5, 6 1036
          XM_017019747.1 XP_016875236.1 5, 6 1039
          XM_017019748.1 XP_016875237.1 6, 7 1300
          XM_017019749.1 XP_016875238.1 6, 7 1064
          XM_017019750.1 XP_016875239.1 6, 7 602
          XM_017019751.1 XP_016875240.1 6, 7 713
          XM_017019752.1 XP_016875241.1 7, 8 811
          XM_017019753.1 XP_016875242.1 6, 7 697
          XM_017019754.1 XP_016875243.1 6, 7 908
          XM_017019755.1 XP_016875244.1 6, 7 1306
          XM_017019756.1 XP_016875245.1 6, 7 602
          XM_017019757.1 XP_016875246.1 7, 8 811
          GenBank mRNA AK225126.1 - 6, 7 587
          AK292256.1 - 7, 8 1131
          AK299105.1 - 6, 7 632
          AK299534.1 - 2, 3 143
          AK301700.1 - 3, 4 348
          AK302742.1 - 6, 7 637
          AK303515.1 - 6, 7 703
          AK312317.1 - 6, 7 1036
          AK316250.1 - 6, 7 587
          BC010621.2 - 6, 7 590 3872822
          KU178292.1 - 5, 6 551
          KU178293.1 - 5, 6 551
          KU178294.1 - 5, 6 551
          U19721.1 - 5, 6 565
          X84899.1 - 5, 6 569
          Z48054.1 - 6, 7 637

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          membrane trafficking regulatory protein

          Gene Ontology Categories:

          Function(s) Process(es)

          protein import into peroxisome matrix
          protein import into peroxisome matrix, docking
          protein import into peroxisome membrane
          negative regulation of protein homotetramerization
          peroxisome targeting sequence binding
          peroxisome matrix targeting signal-1 binding
          protein binding
          enzyme binding
          small GTPase binding
          protein N-terminus binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-2rf75:80/100.66.79.83:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0