Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • Enterprise Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PEX5 SIRNA Assays ›
          Gene Symbol:
          PEX5
          Gene Name:
          peroxisomal biogenesis factor 5
          Gene Aliases:
          PBD2A, PBD2B, PTS1-BP, PTS1R, PXR1, RCDP5
          Chromosome Location:
          Chr.12: 7188072 - 7218574 on Build GRCh38
          UniGene ID:
          Hs.567327
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s532233

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PEX5
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000319.4 NP_000310.2 13 1454
          NM_001131023.1 NP_001124495.1 14 1588
          NM_001131024.1 NP_001124496.1 13 1432
          NM_001131025.1 NP_001124497.1 14 2009
          NM_001131026.1 NP_001124498.1 15 2042
          NM_001300789.1 NP_001287718.1 14 1614
          XM_005253455.1 XP_005253512.1 13 1498
          XM_011520793.2 XP_011519095.1 14 1603
          XM_011520795.1 XP_011519097.1 14 2020
          XM_011520797.1 XP_011519099.1 14 1513
          XM_011520798.1 XP_011519100.1 15 1722
          XM_011520799.2 XP_011519101.1 14 2162
          XM_011520800.1 XP_011519102.1 14 1608
          XM_011520801.2 XP_011519103.1 13 1491
          XM_011520802.2 XP_011519104.1 13 1365
          XM_017019744.1 XP_016875233.1 13 1947
          XM_017019745.1 XP_016875234.1 13 1948
          XM_017019746.1 XP_016875235.1 12 1836
          XM_017019747.1 XP_016875236.1 12 1815
          XM_017019748.1 XP_016875237.1 14 2211
          XM_017019749.1 XP_016875238.1 14 1975
          XM_017019750.1 XP_016875239.1 14 1489
          XM_017019751.1 XP_016875240.1 13 1513
          XM_017019752.1 XP_016875241.1 14 1611
          XM_017019753.1 XP_016875242.1 13 1497
          XM_017019754.1 XP_016875243.1 13 1708
          XM_017019755.1 XP_016875244.1 13 2106
          XM_017019756.1 XP_016875245.1 13 1378
          XM_017019757.1 XP_016875246.1 14 1587
          GenBank mRNA AK225126.1 - 13 1387
          AK292256.1 - 15 2042
          AK299105.1 - 13 1432
          AK299534.1 - 9 943
          AK301700.1 - 10 1148
          AK302742.1 - 14 1548
          AK303515.1 - 14 1614
          AK312317.1 - 14 1947
          AK316250.1 - 13 1387
          BC010621.2 - 14 1477 3872822
          KU178292.1 - 13 1438
          KU178293.1 - 13 1462
          KU178294.1 - 12 1351
          U19721.1 - 12 1365
          X84899.1 - 13 1480
          Z48054.1 - 13 1437

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          membrane trafficking regulatory protein

          Gene Ontology Categories:

          Function(s) Process(es)

          protein import into peroxisome matrix
          protein import into peroxisome matrix, docking
          protein import into peroxisome membrane
          negative regulation of protein homotetramerization
          peroxisome targeting sequence binding
          peroxisome matrix targeting signal-1 binding
          protein binding
          enzyme binding
          small GTPase binding
          protein N-terminus binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0