Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PEX5 SIRNA Assays ›
          Gene Symbol:
          PEX5
          Gene Name:
          peroxisomal biogenesis factor 5
          Gene Aliases:
          PBD2A, PBD2B, PTS1-BP, PTS1R, PXR1, RCDP5
          Chromosome Location:
          Chr.12: 7188072 - 7218574 on Build GRCh38
          UniGene ID:
          Hs.567327
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s532234

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PEX5
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000319.4 NP_000310.2 13, 14 1561
          NM_001131023.1 NP_001124495.1 14, 15 1695
          NM_001131024.1 NP_001124496.1 13, 14 1539
          NM_001131025.1 NP_001124497.1 14, 15 2116
          NM_001131026.1 NP_001124498.1 15, 16 2149
          NM_001300789.1 NP_001287718.1 14, 15 1721
          XM_005253455.1 XP_005253512.1 13, 14 1605
          XM_011520793.2 XP_011519095.1 14, 15 1710
          XM_011520795.1 XP_011519097.1 14, 15 2127
          XM_011520797.1 XP_011519099.1 14, 15 1620
          XM_011520798.1 XP_011519100.1 15, 16 1829
          XM_011520799.2 XP_011519101.1 14, 15 2269
          XM_011520800.1 XP_011519102.1 14, 15 1715
          XM_011520801.2 XP_011519103.1 13, 14 1598
          XM_011520802.2 XP_011519104.1 13, 14 1472
          XM_017019744.1 XP_016875233.1 13, 14 2054
          XM_017019745.1 XP_016875234.1 13, 14 2055
          XM_017019746.1 XP_016875235.1 12, 13 1943
          XM_017019747.1 XP_016875236.1 12, 13 1922
          XM_017019748.1 XP_016875237.1 14, 15 2318
          XM_017019749.1 XP_016875238.1 14, 15 2082
          XM_017019750.1 XP_016875239.1 14, 15 1596
          XM_017019751.1 XP_016875240.1 13, 14 1620
          XM_017019752.1 XP_016875241.1 14, 15 1718
          XM_017019753.1 XP_016875242.1 13, 14 1604
          XM_017019754.1 XP_016875243.1 13, 14 1815
          XM_017019755.1 XP_016875244.1 13, 14 2213
          XM_017019756.1 XP_016875245.1 13, 14 1485
          XM_017019757.1 XP_016875246.1 14, 15 1694
          GenBank mRNA AK225126.1 - 13, 14 1494
          AK292256.1 - 15, 16 2149
          AK299105.1 - 13, 14 1539
          AK299534.1 - 9, 10 1050
          AK301700.1 - 10, 11 1255
          AK302742.1 - 14, 15 1655
          AK303515.1 - 14, 15 1721
          AK312317.1 - 14, 15 2054
          AK316250.1 - 13, 14 1494
          BC010621.2 - 14, 15 1584 3872822
          KU178292.1 - 13, 14 1545
          KU178293.1 - 13, 14 1569
          KU178294.1 - 12, 13 1458
          U19721.1 - 12, 13 1472
          X84899.1 - 13, 14 1587
          Z48054.1 - 13, 14 1544

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          membrane trafficking regulatory protein

          Gene Ontology Categories:

          Function(s) Process(es)

          protein import into peroxisome matrix
          protein import into peroxisome matrix, docking
          protein import into peroxisome membrane
          negative regulation of protein homotetramerization
          peroxisome targeting sequence binding
          peroxisome matrix targeting signal-1 binding
          protein binding
          enzyme binding
          small GTPase binding
          protein N-terminus binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-mc8hj:80/100.66.75.242:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0