Search
Search

ADAMTS17| Gene Symbol: ADAMTS17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139057.3 | NP_620688.2 | 17 | 2437 | ||
| XM_005254872.3 | XP_005254929.1 | 18 | 3434 | |||
| XM_011521312.2 | XP_011519614.1 | 19 | 3503 | |||
| XM_017021973.1 | XP_016877462.1 | 20 | 3636 | |||
| XM_017021974.1 | XP_016877463.1 | 20 | 3638 | |||
| XM_017021975.1 | XP_016877464.1 | 19 | 3567 | |||
| XM_017021976.1 | XP_016877465.1 | 19 | 2348 | |||
| XM_017021978.1 | XP_016877467.1 | 13 | 4894 | |||
| XM_017021979.1 | XP_016877468.1 | 12 | 1344 | |||
| XM_017021980.1 | XP_016877469.1 | 12 | 1554 | |||
| XM_017021982.1 | XP_016877471.1 | 10 | 1164 | |||
| XM_017021983.1 | XP_016877472.1 | 8 | 893 | |||
| GenBank mRNA | AJ315735.1 | - | 17 | 2354 | ||