Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PRKAR1A SIRNA Assays ›
          Gene Symbol:
          PRKAR1A
          Gene Name:
          protein kinase cAMP-dependent type I regulatory subunit alpha
          Gene Aliases:
          ACRDYS1, ADOHR, CAR, CNC, CNC1, PKR1, PPNAD1, PRKAR1, TSE1
          Chromosome Location:
          Chr.17: 68413623 - 68551316 on Build GRCh38
          UniGene ID:
          Hs.280342
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s532365

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PRKAR1A
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001276289.1 NP_001263218.1 11 1234
          NM_001276290.1 NP_001263219.1 9 931
          NM_001278433.1 NP_001265362.1 10 1104
          NM_002734.4 NP_002725.1 10 1101
          NM_212471.2 NP_997636.1 10 1294
          NM_212472.2 NP_997637.1 10 1132
          XM_011524983.2 XP_011523285.1 10 1197
          XM_011524984.2 XP_011523286.1 11 1135
          XM_011524985.2 XP_011523287.1 11 1429
          GenBank mRNA A12295.1 - 10 1027
          AK097580.1 - 9 1504
          AK124586.1 - 10 1020
          AK312307.1 - 10 1030
          BC036285.1 - 10 1053 4340015
          BC093042.1 - 10 1042 6514677
          CR749311.1 - 11 1163
          M18468.1 - 10 1028
          M33336.1 - 10 1012
          S54709.1 - 3 175

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          protein-binding activity modulator kinase modulator

          Gene Ontology Categories:

          Function(s) Process(es)

          mesoderm formation
          renal water homeostasis
          regulation of transcription from RNA polymerase II promoter
          female meiotic division
          blood coagulation
          activation of protein kinase A activity
          intracellular signal transduction
          sarcomere organization
          negative regulation of meiotic nuclear division
          negative regulation of activated T cell proliferation
          cardiac muscle cell proliferation
          cellular response to glucagon stimulus
          negative regulation of cAMP-dependent protein kinase activity
          cAMP-dependent protein kinase inhibitor activity
          protein binding
          cAMP-dependent protein kinase regulator activity
          cAMP binding
          ubiquitin protein ligase binding
          protein kinase A catalytic subunit binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0