Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TSC2 SIRNA Assays ›
          Gene Symbol:
          TSC2
          Gene Name:
          tuberous sclerosis 2
          Gene Aliases:
          LAM, PPP1R160, TSC4
          Chromosome Location:
          Chr.16: 2047804 - 2088720 on Build GRCh38
          UniGene ID:
          Hs.90303
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s533032

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TSC2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000548.4 NP_000539.2 3, 4 417
          NM_001077183.2 NP_001070651.1 3, 4 417
          NM_001114382.2 NP_001107854.1 3, 4 417
          NM_001318829.1 NP_001305758.1 2, 3 250
          NM_001318832.1 NP_001305761.1 3, 4 318
          XM_005255529.4 XP_005255586.2 3, 4 304
          XM_005255531.4 XP_005255588.2 3, 4 304
          XM_011522636.2 XP_011520938.1 3, 4 304
          XM_011522637.2 XP_011520939.1 3, 4 304
          XM_011522639.2 XP_011520941.1 3, 4 304
          XM_011522640.2 XP_011520942.1 3, 4 304
          XM_017023615.1 XP_016879104.1 3, 4 304
          XM_017023616.1 XP_016879105.1 3, 4 304
          XM_017023618.1 XP_016879107.1 3, 4 304
          GenBank mRNA AB210000.1 - 3, 4 323
          AK294548.1 - 3, 4 315
          AK295672.1 - 2, 3 137
          AK295728.1 - 3, 4 318
          BC150300.1 - 3, 4 292 8860381
          DA316794.1 - 3, 4 332
          DC402106.1 - 3, 4 419
          KJ534954.1 - 2, 3 217
          KJ534955.1 - 2, 3 217
          KJ534956.1 - 2, 3 217
          KJ534957.1 - 2, 3 217
          KJ534958.1 - 2, 3 217
          KJ534982.1 - 2, 3 217
          KJ534983.1 - 2, 3 217
          KJ534984.1 - 2, 3 217
          KJ534985.1 - 2, 3 217
          KJ535011.1 - 2, 3 217
          KJ535022.1 - 2, 3 217
          KJ535038.1 - 2, 3 217
          KJ535039.1 - 2, 3 217
          KJ535051.1 - 2, 3 217
          KJ535091.1 - 2, 3 217
          X75621.1 - 2, 3 235

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          GTPase-activating protein

          Gene Ontology Categories:

          Function(s) Process(es)

          neural tube closure
          negative regulation of protein kinase activity
          protein import into nucleus
          endocytosis
          cell cycle arrest
          heart development
          protein localization
          negative regulation of cell proliferation
          negative regulation of phosphatidylinositol 3-kinase signaling
          viral process
          vesicle-mediated transport
          regulation of endocytosis
          negative regulation of Wnt signaling pathway
          negative regulation of TOR signaling
          protein kinase B signaling
          positive regulation of GTPase activity
          regulation of insulin receptor signaling pathway
          negative regulation of insulin receptor signaling pathway
          insulin-like growth factor receptor signaling pathway
          positive chemotaxis
          regulation of small GTPase mediated signal transduction
          regulation of cell cycle
          negative regulation of protein kinase B signaling
          GTPase activator activity
          protein binding
          phosphatase binding
          small GTPase binding
          protein homodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0