Search
Search

GALNT14| Gene Symbol: GALNT14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001253826.1 | NP_001240755.1 | 13 | 1875 | ||
| NM_001253827.1 | NP_001240756.1 | 14 | 1640 | |||
| NM_024572.3 | NP_078848.2 | 12 | 1860 | |||
| XM_011533105.1 | XP_011531407.1 | 14 | 1394 | |||
| XM_017004906.1 | XP_016860395.1 | 12 | 1325 | |||
| GenBank mRNA | AB078144.1 | - | 12 | 1220 | ||
| AJ505966.1 | - | 1 | 77 | |||
| AK022753.1 | - | 13 | 1296 | |||
| AK074394.1 | - | 6 | 520 | |||
| AK091313.1 | - | 12 | 1230 | |||
| AK122747.1 | - | 14 | 1624 | |||
| AK225808.1 | - | 12 | 1583 | |||
| AK298735.1 | - | 13 | 1288 | |||
| AY358758.1 | - | 12 | 1553 | |||
| BC006269.2 | - | 5 | 429 | |||
| BC010659.1 | - | 12 | 1860 | 3874588 | ||
| Y09324.4 | - | 12 | 1220 | |||