Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • Enterprise Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TCF4 SIRNA Assays ›
          Gene Symbol:
          TCF4
          Gene Name:
          transcription factor 4
          Gene Aliases:
          E2-2, FECD3, ITF-2, ITF2, PTHS, SEF-2, SEF2, SEF2-1, SEF2-1A, SEF2-1B, SEF2-1D, TCF-4, bHLHb19
          Chromosome Location:
          Chr.18: 55222331 - 55635993 on Build GRCh38
          UniGene ID:
          Hs.605153
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s533402

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 392.00
          Your Price
          Online offer: 380.65
          380.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TCF4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001083962.1 NP_001077431.1 12, 13 1599
          NM_001243226.2 NP_001230155.2 13, 14 1389
          NM_001243227.1 NP_001230156.1 11, 12 1433
          NM_001243228.1 NP_001230157.1 12, 13 1543
          NM_001243230.1 NP_001230159.1 11, 12 1075
          NM_001243231.1 NP_001230160.1 10, 11 1033
          NM_001243232.1 NP_001230161.1 8, 9 1055
          NM_001243233.1 NP_001230162.1 8, 9 996
          NM_001243234.1 NP_001230163.1 5, 6 628
          NM_001243235.1 NP_001230164.1 5, 6 628
          NM_001243236.1 NP_001230165.1 5, 6 644
          NM_001306207.1 NP_001293136.1 11, 12 1224
          NM_001306208.1 NP_001293137.1 8, 9 1055
          NM_003199.2 NP_003190.1 12, 13 1599
          XM_005266739.3 XP_005266796.2 11, 12 1239
          XM_005266741.3 XP_005266798.1 12, 13 1199
          XM_005266743.4 XP_005266800.1 11, 12 1060
          XM_005266744.3 XP_005266801.1 11, 12 1180
          XM_005266745.3 XP_005266802.1 11, 12 1042
          XM_005266747.3 XP_005266804.1 10, 11 1036
          XM_005266749.3 XP_005266806.1 8, 9 827
          XM_005266752.4 XP_005266809.1 8, 9 1397
          XM_005266754.3 XP_005266811.1 7, 8 821
          XM_005266755.4 XP_005266812.1 8, 9 856
          XM_005266761.3 XP_005266818.1 5, 6 1140
          XM_006722536.2 XP_006722599.1 12, 13 1522
          XM_006722537.2 XP_006722600.1 12, 13 1155
          XM_006722538.2 XP_006722601.1 11, 12 1177
          XM_011526158.2 XP_011524460.1 11, 12 2153
          XM_011526160.2 XP_011524462.1 7, 8 1206
          XM_017025934.1 XP_016881423.1 11, 12 1236
          XM_017025935.1 XP_016881424.1 11, 12 1239
          XM_017025936.1 XP_016881425.1 11, 12 1239
          XM_017025937.1 XP_016881426.1 11, 12 993
          XM_017025938.1 XP_016881427.1 12, 13 1152
          XM_017025939.1 XP_016881428.1 12, 13 1202
          XM_017025940.1 XP_016881429.1 12, 13 1522
          XM_017025941.1 XP_016881430.1 12, 13 1155
          XM_017025942.1 XP_016881431.1 12, 13 1199
          XM_017025943.1 XP_016881432.1 12, 13 1202
          XM_017025944.1 XP_016881433.1 11, 12 1457
          XM_017025945.1 XP_016881434.1 11, 12 1460
          XM_017025946.1 XP_016881435.1 11, 12 1474
          XM_017025947.1 XP_016881436.1 11, 12 1060
          XM_017025948.1 XP_016881437.1 11, 12 1457
          XM_017025949.1 XP_016881438.1 11, 12 1177
          XM_017025950.1 XP_016881439.1 11, 12 1457
          XM_017025951.1 XP_016881440.1 10, 11 1036
          XM_017025952.1 XP_016881441.1 8, 9 1050
          XM_017025953.1 XP_016881442.1 8, 9 827
          XM_017025954.1 XP_016881443.1 8, 9 1411
          XM_017025955.1 XP_016881444.1 7, 8 1145
          XM_017025956.1 XP_016881445.1 7, 8 821
          XM_017025957.1 XP_016881446.1 5, 6 1140
          GenBank mRNA AB209741.1 - 7, 8 707
          AK095041.1 - 8, 9 1055
          AK096862.1 - 10, 11 1033
          AK122765.1 - 12, 13 1599
          AK299169.1 - 11, 12 1433
          AK300038.1 - 5, 6 574
          AK300612.1 - 8, 9 996
          AK300636.1 - 7, 8 726
          AK301144.1 - 11, 12 1210
          AK315074.1 - 12, 13 1531
          AK316165.1 - 7, 8 770
          AL518903.3 - 5, 6 644
          BC031056.1 - 13, 14 1350
          BC125084.1 - 11, 12 1013 40117795
          BC125085.1 - 11, 12 1013 40117798
          FR748210.1 - 11, 12 987
          FR748211.1 - 11, 12 987
          FR748212.1 - 9, 10 807
          FR748213.1 - 9, 10 807
          FR748214.1 - 10, 11 915
          FR748215.1 - 8, 9 735
          FR748216.1 - 11, 12 981
          FR748217.1 - 10, 11 861
          FR748218.1 - 6, 7 597
          FR748219.1 - 8, 9 774
          FR748220.1 - 5, 6 507
          FR748221.1 - 5, 6 507
          FR748222.1 - 5, 6 507
          FR748223.1 - 3, 4 339
          M74718.1 - 5, 6 459
          M74719.1 - 12, 13 1186
          X52079.1 - 9, 10 852

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          basic helix-loop-helix transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA-templated transcription, initiation
          transcription initiation from RNA polymerase II promoter
          positive regulation of neuron differentiation
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          protein-DNA complex assembly
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          transcription factor activity, sequence-specific DNA binding, RNA polymerase recruiting
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          transcription factor activity, TFIIB-class binding
          TFIIB-class transcription factor binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          protein binding
          protein C-terminus binding
          identical protein binding
          protein homodimerization activity
          bHLH transcription factor binding
          protein heterodimerization activity
          E-box binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-2rf75:80/100.66.79.83:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0