Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CLDN16 SIRNA Assays ›
          Gene Symbol:
          CLDN16
          Gene Name:
          claudin 16
          Gene Aliases:
          HOMG3, PCLN1
          Chromosome Location:
          Chr.3: 190387872 - 190412143 on Build GRCh38
          UniGene ID:
          Hs.251391
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s533450

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CLDN16
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_006580.3 NP_006571.1 1 484
          GenBank mRNA AF152101.1 - 1 304
          AK314955.1 - 1 483
          BC069662.1 - 1 304 7262563
          BC069682.1 - 1 304 7262480
          BC069759.1 - 1 304 7262492
          BC069777.1 - 1 304 7262504
          DQ305102.1 - 1 236
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA DA621288.1 - 1 203 Chr. 3 - 190388154 - 190404931 [+] on Build GRCh38
          GenBankmRNA BG433757.1 - 149 Chr. - - [-] on Build
          GenBankmRNA CD622944.1 - 1 178 Chr. 3 - 190388177 - 190404860 [+] on Build GRCh38
          GenBankmRNA DA636744.1 - 1 194 Chr. 3 - 190388163 - 190404874 [+] on Build GRCh38
          GenBankmRNA CD622942.1 - 1 177 Chr. 3 - 190388179 - 190404860 [+] on Build GRCh38
          GenBankmRNA DA539435.1 - 1 117 Chr. 3 - 190388239 - 190408418 [+] on Build GRCh38
          GenBankmRNA DA623344.1 - 1 205 Chr. 3 - 190388152 - 190404904 [+] on Build GRCh38
          GenBankmRNA CD622948.1 - 1 150 Chr. 3 - 190388210 - 190404860 [+] on Build GRCh38
          GenBankmRNA BP274478.1 - 1 202 Chr. 3 - 190388154 - 190408335 [+] on Build GRCh38
          GenBankmRNA CD622947.1 - 1 128 Chr. 3 - 190388227 - 190404860 [+] on Build GRCh38
          GenBankmRNA BP276177.1 - 1 106 Chr. 3 - 190388250 - 190408382 [+] on Build GRCh38
          GenBankmRNA EL736324.1 - 1 236 Chr. 3 - 190388120 - 190408329 [+] on Build GRCh38
          GenBankmRNA DA627119.1 - 1 483 Chr. 3 - 190387872 - 190388410 [+] on Build GRCh38

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          tight junction

          Gene Ontology Categories:

          Function(s) Process(es)

          ion transport
          cellular metal ion homeostasis
          excretion
          calcium-independent cell-cell adhesion via plasma membrane cell-adhesion molecules
          positive regulation of calcium ion transport
          transepithelial transport
          magnesium ion transmembrane transport
          structural molecule activity
          protein binding
          magnesium ion transmembrane transporter activity
          identical protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0