Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other DICER1 SIRNA Assays ›
          Gene Symbol:
          DICER1
          Gene Name:
          dicer 1, ribonuclease III
          Gene Aliases:
          DCR1, Dicer, Dicer1e, HERNA, K12H4.8-LIKE, MNG1, RMSE2
          Chromosome Location:
          Chr.14: 95086228 - 95158010 on Build GRCh38
          UniGene ID:
          Hs.738957
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s533675

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: DICER1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001195573.1 NP_001182502.1 21 4156
          NM_001271282.2 NP_001258211.1 22 4344
          NM_001291628.1 NP_001278557.1 22 4232
          NM_030621.4 NP_085124.2 24 4403
          NM_177438.2 NP_803187.1 22 4349
          XM_011536599.1 XP_011534901.1 24 4535
          XM_011536600.2 XP_011534902.1 24 4410
          XM_011536601.2 XP_011534903.1 25 4555
          XM_011536602.2 XP_011534904.1 25 4446
          XM_011536604.2 XP_011534906.1 21 4231
          XM_011536605.2 XP_011534907.1 15 9036
          XM_017021120.1 XP_016876609.1 24 4842
          XM_017021121.1 XP_016876610.1 22 4237
          XM_017021122.1 XP_016876611.1 23 4281
          XM_017021123.1 XP_016876612.1 23 4712
          GenBank mRNA AB023145.2 - 22 4349
          AB028449.1 - 23 4299
          AJ132261.2 - 22 4352
          AK001827.1 - 2 146
          AK002007.1 - 1 72
          AK091513.1 - 5 1089
          BC150287.1 - 22 4349 8860125
          HM595745.1 - 21 4120
          KJ175111.1 - 3 1098

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          apoptotic DNA fragmentation
          negative regulation of Schwann cell proliferation
          positive regulation of Schwann cell differentiation
          nerve development
          production of siRNA involved in RNA interference
          targeting of mRNA for destruction involved in RNA interference
          gene silencing by RNA
          pre-miRNA processing
          positive regulation of myelination
          peripheral nervous system myelin formation
          conversion of ds siRNA to ss siRNA involved in RNA interference
          siRNA loading onto RISC involved in RNA interference
          production of miRNAs involved in gene silencing by miRNA
          miRNA loading onto RISC involved in gene silencing by miRNA
          conversion of ds siRNA to ss siRNA
          neuron projection morphogenesis
          RNA phosphodiester bond hydrolysis, endonucleolytic
          double-stranded RNA binding
          helicase activity
          endoribonuclease activity
          ribonuclease III activity
          deoxyribonuclease I activity
          protein binding
          ATP binding
          protein domain specific binding
          siRNA binding
          metal ion binding
          pre-miRNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-srbv6:80/100.66.75.104:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0