Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: B9D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243473.2 | NP_001230402.1 | 5 | 466 | ||
NM_001243475.2 | NP_001230404.1 | 5 | 662 | |||
NM_001321214.1 | NP_001308143.1 | 5 | 559 | |||
NM_001321215.1 | NP_001308144.1 | 5 | 559 | |||
NM_001321216.1 | NP_001308145.1 | 5 | 559 | |||
NM_001321217.1 | NP_001308146.1 | 5 | 559 | |||
NM_001321218.1 | NP_001308147.1 | 5 | 559 | |||
NM_001321219.1 | NP_001308148.1 | 5 | 559 | |||
NM_015681.4 | NP_056496.1 | 5 | 559 | |||
XM_005256607.2 | XP_005256664.1 | 5 | 531 | |||
XM_005256610.1 | XP_005256667.1 | 5 | 534 | |||
XM_017024452.1 | XP_016879941.1 | 8 | 2923 | |||
GenBank mRNA | AB030506.1 | - | 5 | 548 | ||
AK293815.1 | - | 5 | 532 | |||
AK297782.1 | - | 5 | 449 | |||
AK309320.1 | - | 5 | 651 | |||
BC002944.2 | - | 5 | 490 | 3953287 | ||
BI833202.1 | - | 5 | 554 |