Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other LIG4 SIRNA Assays ›
          Gene Symbol:
          LIG4
          Gene Name:
          DNA ligase 4
          Gene Aliases:
          LIG4S
          Chromosome Location:
          Chr.13: 108207442 - 108218368 on Build GRCh38
          UniGene ID:
          Hs.166091
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s534351

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: LIG4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001098268.1 NP_001091738.1 2 798
          NM_002312.3 NP_002303.2 2 836
          NM_206937.1 NP_996820.1 3 715
          XM_005254056.1 XP_005254113.1 3 680
          XM_005254057.4 XP_005254114.1 3 850
          XM_005254058.3 XP_005254115.1 2 2296
          XM_006719951.3 XP_006720014.1 3 2555
          XM_006719952.1 XP_006720015.1 4 1148
          XM_011521091.2 XP_011519393.1 4 1177
          XM_011521092.2 XP_011519394.1 3 753
          XM_017020563.1 XP_016876052.1 3 2397
          XM_017020564.1 XP_016876053.1 2 823
          XM_017020565.1 XP_016876054.1 4 1194
          XM_017020566.1 XP_016876055.1 4 1097
          XM_017020567.1 XP_016876056.1 3 751
          XM_017020568.1 XP_016876057.1 3 2373
          XM_017020569.1 XP_016876058.1 4 2818
          XM_017020570.1 XP_016876059.1 4 1097
          XM_017020571.1 XP_016876060.1 4 2902
          XM_017020572.1 XP_016876061.1 3 2289
          XM_017020573.1 XP_016876062.1 2 599
          GenBank mRNA AK292419.1 - 3 624
          AK314844.1 - 3 680
          BC037491.2 - 3 715 5259632
          BX648833.1 - 2 798
          X83441.1 - 2 836

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          DNA metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          single strand break repair
          in utero embryonic development
          pro-B cell differentiation
          DNA ligation
          lagging strand elongation
          nucleotide-excision repair, DNA gap filling
          double-strand break repair
          double-strand break repair via nonhomologous end joining
          cell cycle
          central nervous system development
          cell proliferation
          response to X-ray
          response to gamma radiation
          T cell differentiation in thymus
          V(D)J recombination
          immunoglobulin V(D)J recombination
          T cell receptor V(D)J recombination
          somatic stem cell population maintenance
          negative regulation of neuron apoptotic process
          isotype switching
          positive regulation of fibroblast proliferation
          positive regulation of neurogenesis
          DNA ligation involved in DNA recombination
          DNA ligation involved in DNA repair
          chromosome organization
          cell division
          neuron apoptotic process
          cellular response to lithium ion
          DNA biosynthetic process
          establishment of integrated proviral latency
          double-strand break repair via classical nonhomologous end joining
          positive regulation of chromosome organization
          DNA binding
          DNA ligase activity
          DNA ligase (ATP) activity
          protein binding
          ATP binding
          protein C-terminus binding
          ligase activity
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0