Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: KHDC3L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017361.2 | NP_001017361.1 | 1 | 175 | ||
GenBank mRNA | AB211062.1 | - | 1 | 175 | ||
BC132844.1 | - | 1 | 167 | 40146866 | ||
BC137160.1 | - | 1 | 167 | 9021157 |