Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: FANCE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021922.2 | NP_068741.1 | 2 | 676 | ||
XM_005248885.2 | XP_005248942.1 | 2 | 698 | |||
XM_005248886.2 | XP_005248943.1 | 2 | 698 | |||
XM_005248887.2 | XP_005248944.1 | 2 | 698 | |||
XM_005248888.3 | XP_005248945.1 | 2 | 698 | |||
XM_011514343.2 | XP_011512645.1 | 2 | 391 | |||
GenBank mRNA | AF265210.1 | - | 2 | 676 | ||
AK292522.1 | - | 2 | 687 | |||
AK312617.1 | - | 2 | 664 | |||
BC046359.1 | - | 2 | 682 | 6154799 |