Search
Search

IGSF1| Gene Symbol: IGSF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170961.1 | NP_001164432.1 | 5 | 848 | ||
| NM_001170962.1 | NP_001164433.1 | 4 | 821 | |||
| NM_001170963.1 | NP_001164434.1 | 5 | 672 | |||
| NM_001555.4 | NP_001546.2 | 5 | 848 | |||
| NM_205833.3 | NP_991402.1 | 5 | 848 | |||
| XM_011531330.1 | XP_011529632.1 | 6 | 746 | |||
| XM_011531333.1 | XP_011529635.1 | 5 | 848 | |||
| XM_011531334.1 | XP_011529636.1 | 5 | 848 | |||
| GenBank mRNA | AB058894.2 | - | 5 | 700 | ||
| AF034198.1 | - | 4 | 670 | |||
| AK057916.1 | - | 5 | 679 | |||
| AK226008.1 | - | 5 | 705 | |||
| AK307871.1 | - | 5 | 724 | |||
| BC063884.1 | - | 5 | 684 | 30387876 | ||
| Y10523.1 | - | 4 | 645 | |||