Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PLA2G6 SIRNA Assays ›
          Gene Symbol:
          PLA2G6
          Gene Name:
          phospholipase A2 group VI
          Gene Aliases:
          CaI-PLA2, GVI, INAD1, IPLA2-VIA, NBIA2, NBIA2A, NBIA2B, PARK14, PLA2, PNPLA9, iPLA2, iPLA2beta
          Chromosome Location:
          Chr.22: 38111495 - 38192099 on Build GRCh38
          UniGene ID:
          Hs.170479
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s536327

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 392.00
          Your Price
          Online offer: 380.65
          380.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PLA2G6
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001004426.1 NP_001004426.1 16 2682
          NM_001199562.1 NP_001186491.1 16 2637
          NM_003560.2 NP_003551.2 17 2844
          XM_005261764.2 XP_005261821.1 17 2819
          XM_005261765.1 XP_005261822.1 17 2816
          XM_005261766.1 XP_005261823.1 17 2911
          XM_006724332.3 XP_006724395.1 17 2820
          XM_011530422.1 XP_011528724.1 17 2695
          XM_011530423.1 XP_011528725.1 18 2931
          XM_011530424.1 XP_011528726.1 19 3055
          XM_011530425.1 XP_011528727.1 17 2755
          XM_017028981.1 XP_016884470.1 16 2749
          XM_017028982.1 XP_016884471.1 16 2654
          XM_017028983.1 XP_016884472.1 18 2893
          XM_017028984.1 XP_016884473.1 17 2769
          XM_017028985.1 XP_016884474.1 16 2587
          GenBank mRNA AF064594.1 - 17 2844
          AF102988.1 - 15 2546
          AF102989.1 - 16 2708
          AK096521.1 - 16 2377
          AK128862.1 - 6 3595
          AL080187.1 - 17 2839
          BC036742.1 - 17 2833 5166749
          BC051904.1 - 16 2618 5175768

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          phospholipase

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of protein phosphorylation
          chemotaxis
          positive regulation of cytosolic calcium ion concentration
          memory
          urinary bladder smooth muscle contraction
          lipid catabolic process
          antibacterial humoral response
          cardiolipin biosynthetic process
          response to endoplasmic reticulum stress
          positive regulation of insulin secretion involved in cellular response to glucose stimulus
          phosphatidylcholine acyl-chain remodeling
          phosphatidylethanolamine acyl-chain remodeling
          Fc-gamma receptor signaling pathway involved in phagocytosis
          positive regulation of vasodilation
          positive regulation of exocytosis
          negative regulation of synaptic transmission, glutamatergic
          maternal process involved in female pregnancy
          positive regulation of protein kinase C signaling
          positive regulation of release of cytochrome c from mitochondria
          positive regulation of arachidonic acid secretion
          regulation of store-operated calcium channel activity
          positive regulation of ceramide biosynthetic process
          phospholipase A2 activity
          calmodulin binding
          hydrolase activity
          protein kinase binding
          ATP-dependent protein binding
          calcium-independent phospholipase A2 activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0