Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MECP2 SIRNA Assays ›
          Gene Symbol:
          MECP2
          Gene Name:
          methyl-CpG binding protein 2
          Gene Aliases:
          AUTSX3, MRX16, MRX79, MRXS13, MRXSL, PPMX, RS, RTS, RTT
          Chromosome Location:
          Chr.X: 154021800 - 154097731 on Build GRCh38
          UniGene ID:
          Hs.200716
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s536502

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: MECP2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001110792.1 NP_001104262.1 2 418
          NM_001316337.1 NP_001303266.1 4 710
          NM_004992.3 NP_004983.1 3 542
          XM_005274682.4 XP_005274739.1 4 616
          XM_006724819.3 XP_006724882.1 2 402
          XM_011531166.2 XP_011529468.1 2 429
          XM_017029533.1 XP_016885022.1 6 1042
          GenBank mRNA AF158180.1 - 3 483
          AJ132917.1 - 2 391
          AK289444.1 - 3 517
          AY541280.1 - 2 359
          BC011612.1 - 2 400 3956518
          BI767019.1 - 4 640
          BX538060.1 - 2 408
          GQ203293.1 - 3 542
          GQ203294.1 - 3 542
          GQ203295.1 - 3 542
          GQ896382.1 - 3 542
          GU479943.1 - 3 542
          GU812285.1 - 2 316
          GU812286.1 - 2 316
          HM020402.1 - 3 542
          HM156733.1 - 3 542
          HQ127345.1 - 2 316
          HQ141377.1 - 2 316
          HQ141378.1 - 3 542
          HQ154629.1 - 3 542
          KU178174.1 - 2 316
          KU178175.1 - 2 316
          KU178176.1 - 2 316
          L37298.1 - 2 397
          X89430.1 - 1 287
          X99686.1 - 2 390
          Y12643.1 - 2 380

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          DNA metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          behavioral fear response
          response to hypoxia
          startle response
          neurological system process involved in regulation of systemic arterial blood pressure
          regulation of respiratory gaseous exchange by neurological system process
          inositol metabolic process
          mitochondrial electron transport, ubiquinol to cytochrome c
          chromatin silencing
          regulation of gene expression by genetic imprinting
          transcription, DNA-templated
          glutamine metabolic process
          cellular biogenic amine metabolic process
          synapse assembly
          heart development
          respiratory gaseous exchange
          long-term memory
          locomotory behavior
          protein localization
          glucocorticoid metabolic process
          positive regulation of cell proliferation
          adult locomotory behavior
          visual learning
          response to radiation
          pathogenesis
          post-embryonic development
          response to ionizing radiation
          response to lead ion
          dendrite development
          histone methylation
          histone acetylation
          proprioception
          sensory perception of pain
          cerebellum development
          ventricular system development
          negative regulation of histone methylation
          cardiolipin metabolic process
          response to estradiol
          negative regulation of histone acetylation
          social behavior
          neuron maturation
          negative regulation of neuron apoptotic process
          regulation of DNA methylation
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          phosphatidylcholine metabolic process
          negative regulation of astrocyte differentiation
          catecholamine secretion
          positive regulation of synapse assembly
          excitatory postsynaptic potential
          long-term synaptic potentiation
          four-way junction DNA binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          transcription corepressor activity
          mRNA binding
          protein binding
          transcription factor binding
          methyl-CpG binding
          double-stranded methylated DNA binding
          protein domain specific binding
          chromatin DNA binding
          siRNA binding
          histone deacetylase binding
          poly(A) RNA binding
          unmethylated CpG binding
          protein N-terminus binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-2rf75:80/100.66.79.83:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0