Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other GBA SIRNA Assays ›
          Gene Symbol:
          GBA GBAP1
          Gene Name:
          glucosylceramidase beta
          glucosylceramidase beta pseudogene 1
          Gene Aliases:
          GBA1, GCB, GLUC
          GBAP
          Chromosome Location:
          Chr.1: 155234448 - 155244862 on Build GRCh38
          Chromosome Location:
          Chr.1: 155213825 - 155227534 on Build GRCh38
          UniGene ID:
          Hs.282997
          Hs.719930
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID s536662

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: GBA
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000157.3 NP_000148.2 9 1485
          NM_001005741.2 NP_001005741.1 10 1744
          NM_001005742.2 NP_001005742.1 10 1725
          NM_001171811.1 NP_001165282.1 8 1561
          NM_001171812.1 NP_001165283.1 8 1335
          GenBank mRNA AK291911.1 - 10 1723
          AK298900.1 - 8 1335
          AK300186.1 - 8 1405
          AK300829.1 - 8 1398
          AK300876.1 - 10 1725
          AK301374.1 - 7 1094
          AK302000.1 - 8 1310
          AK312502.1 - 10 1541
          BC003356.1 - 9 1424 2899915
          BX648487.1 - 10 1553
          D13286.1 - 9 1424
          K02920.1 - 8 1304
          KJ690771.1 - 9 1368
          M16328.1 - 9 1754
          M19285.1 - 9 1441
          Gene Symbol: GBAP1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA KJ690774.1 - 1119
          KJ690776.1 - 1009

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          carbohydrate metabolic process
          glucosylceramide catabolic process
          glycosphingolipid metabolic process
          cellular response to starvation
          response to pH
          positive regulation of macroautophagy
          regulation of macroautophagy
          termination of signal transduction
          regulation of proteasomal ubiquitin-dependent protein catabolic process
          negative regulation of interleukin-6 production
          regulation of water loss via skin
          response to testosterone
          positive regulation of protein dephosphorylation
          negative regulation of MAP kinase activity
          skin morphogenesis
          response to estrogen
          sphingosine biosynthetic process
          ceramide biosynthetic process
          negative regulation of inflammatory response
          response to glucocorticoid
          cellular response to tumor necrosis factor
          response to thyroid hormone
          positive regulation of proteolysis involved in cellular protein catabolic process
          glucosylceramidase activity
          receptor binding
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0