Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other FTH1 SIRNA Assays ›
          Gene Symbol:
          FTH1 FTH1P3 FTH1P5
          Gene Name:
          ferritin heavy chain 1
          ferritin heavy chain 1 pseudogene 3
          ferritin heavy chain 1 pseudogene 5
          Gene Aliases:
          FHC, FTH, FTHL6, HFE5, PIG15, PLIF
          FTHL3, FTHL3P
          FTHL5, FTHP1
          Chromosome Location:
          Chr.11: 61964285 - 61967660 on Build GRCh38
          Chromosome Location:
          Chr.2: 27392623 - 27393576 on Build GRCh38
          Chromosome Location:
          Chr.6: 50912550 - 50913462 on Build GRCh38
          UniGene ID:
          Hs.524910
          Hs.658438
          Hs.453583
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          siRNA ID s5383

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: FTH1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_002032.2 NP_002023.2 2, 3 489
          GenBank mRNA AF088851.1 - 2, 3 296
          AK311818.1 - 2, 3 460
          AY258285.1 - 1047
          BC000857.1 - 2, 3 448 3459353
          BC001399.2 - 2, 3 417 3051458
          BC011359.2 - 2, 3 439 4157296
          BC013724.2 - 2, 3 442 3857790
          BC015156.1 - 2, 3 312 3883694
          BC016009.1 - 2, 3 464 4666545
          BC016857.1 - 2, 3 408 3848510
          BC020300.1 - 2, 3 425
          BC035441.1 - 2, 3 448
          BC063514.1 - 2, 3 431 3882345
          BC066961.1 - 2, 3 462 4796312
          BC070494.1 - 3, 4 403
          BC073750.1 - 2, 3 446 6256153
          BC104643.1 - 2, 3 464 4043391
          BC105802.1 - 3, 4 441
          BQ887222.1 - 489
          L20941.1 - 2, 3 462
          M11146.1 - 2, 3 331
          M12937.1 - 2, 3 280
          M97164.1 - 2, 3 383
          X00318.1 - 2, 3 345
          Gene Symbol: FTH1P3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_002201.1 1 463
          GenBank mRNA BC073777.1 - 1 474
          Gene Symbol: FTH1P5
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA J04755.1 - 1 1378

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          storage protein

          Gene Ontology Categories:

          Function(s) Process(es)

          iron ion transport
          cellular iron ion homeostasis
          intracellular sequestering of iron ion
          immune response
          negative regulation of cell proliferation
          negative regulation of fibroblast proliferation
          oxidation-reduction process
          ferroxidase activity
          iron ion binding
          protein binding
          ferric iron binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8lnnk:80/100.66.76.235:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0