Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other AGAP6 SIRNA Assays ›
          Gene Symbol:
          AGAP6 AGAP12P AGAP4 AGAP5 AGAP7P AGAP9 LOC107984026
          Gene Name:
          ArfGAP with GTPase domain, ankyrin repeat and PH domain 6
          ArfGAP with GTPase domain, ankyrin repeat and PH domain 12, pseudogene
          ArfGAP with GTPase domain, ankyrin repeat and PH domain 4
          ArfGAP with GTPase domain, ankyrin repeat and PH domain 5
          ArfGAP with GTPase domain, ankyrin repeat and PH domain 7, pseudogene
          ArfGAP with GTPase domain, ankyrin repeat and PH domain 9
          arf-GAP with GTPase, ANK repeat and PH domain-containing protein 4-like
          Gene Aliases:
          AGAP-6, CTGLF3, bA324H6.1
          CTGLF12P
          AGAP-4, AGAP-8, AGAP8, CTGLF1, CTGLF5, MRIP2
          CTGLF2
          AGAP-7, AGAP7, CTGLF4, bA109G10.1
          AGAP-9, CTGLF6, bA301J7.2
          -
          Chromosome Location:
          Chr.10: 49981196 - 50010507 on Build GRCh38
          Chromosome Location:
          Chr.10: 48010132 - 48031640 on Build GRCh38
          Chromosome Location:
          Chr.10: 45825592 - 45858529 on Build GRCh38
          Chromosome Location:
          Chr.10: 73674275 - 73697796 on Build GRCh38
          Chromosome Location:
          Chr.10: 46109495 - 46131678 on Build GRCh38
          Chromosome Location:
          Chr.10: 47501854 - 47523638 on Build GRCh38
          Chromosome Location:
          Chr.10: 46335552 - 46838268 on Build GRCh38
          UniGene ID:
          Hs.536158
          Hs.645213
          Hs.314437
          Hs.651147
          Hs.447454
          Hs.645346
          -
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          -
          siRNA ID s53913

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: AGAP6
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001077665.2 NP_001071133.2 8 1626
          XM_005269841.3 XP_005269898.1 2 667
          XM_011539814.2 XP_011538116.1 2 671
          XM_011539815.2 XP_011538117.1 5 851
          XM_017016268.1 XP_016871757.1 8 1347
          XM_017016269.1 XP_016871758.1 8 1347
          XM_017016270.1 XP_016871759.1 7 1581
          XM_017016271.1 XP_016871760.1 10 1349
          XM_017016272.1 XP_016871761.1 9 1279
          XM_017016273.1 XP_016871762.1 9 1972
          XM_017016274.1 XP_016871763.1 5 802
          Gene Symbol: AGAP12P
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_029396.2 8 1270
          Gene Symbol: AGAP4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001276343.2 NP_001263272.2 8 1354
          NM_001291379.1 NP_001278308.1 10 1546
          NM_133446.3 NP_597703.2 7 1285
          XM_011539269.2 XP_011537571.1 6 892
          XM_017015664.1 XP_016871153.1 10 5216
          XM_017015665.1 XP_016871154.1 11 5084
          XM_017015666.1 XP_016871155.1 9 2119
          XM_017015667.1 XP_016871156.1 10 1389
          XM_017015668.1 XP_016871157.1 8 1139
          XM_017015669.1 XP_016871158.1 8 1047
          GenBank mRNA AF411132.1 - 7 1285
          BC048341.1 - 1 407
          BC171820.1 - 8 1339
          Gene Symbol: AGAP5
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001144000.1 NP_001137472.1 8 1269
          Gene Symbol: AGAP7P
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_126580.1 7 1254
          GenBank mRNA BY996334.1 - 1 319
          Gene Symbol: AGAP9
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001190810.1 NP_001177739.1 8 1340
          GenBank mRNA AK160375.1 - 9 4316
          BC171845.1 - 8 1340
          Gene Symbol: LOC107984026
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_017017029.1 XP_016872518.1 10 1771
          XM_017017030.1 XP_016872519.1 9 1702
          XM_017017031.1 XP_016872520.1 9 1702
          XM_017017032.1 XP_016872521.1 11 1802
          XM_017017033.1 XP_016872522.1 12 1565
          XM_017017034.1 XP_016872523.1 11 1470
          XM_017017035.1 XP_016872524.1 7 983
          XM_017017036.1 XP_016872525.1 5 1144
          XM_017017037.1 XP_016872526.1 7 1533
          XM_017017038.1 XP_016872527.1 5 786
          XM_017017039.1 XP_016872528.1 7 1002
          XM_017017040.1 XP_016872529.1 7 966

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of GTPase activity
          GTPase activator activity
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-77fzl:80/100.66.72.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0