Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PRAMEF6 SIRNA Assays ›
          Gene Symbol:
          PRAMEF6 PRAMEF11 PRAMEF15 PRAMEF25 PRAMEF26 PRAMEF27 PRAMEF4 PRAMEF5 PRAMEF9
          Gene Name:
          PRAME family member 6
          PRAME family member 11
          PRAME family member 15
          PRAME family member 25
          PRAME family member 26
          PRAME family member 27
          PRAME family member 4
          PRAME family member 5
          PRAME family member 9
          Gene Aliases:
          -
          -
          -
          -
          -
          -
          -
          PRAMEF23, PRAMEF5L
          -
          Chromosome Location:
          Chr.1: 12938472 - 12947580 on Build GRCh38
          Chromosome Location:
          Chr.1: 12824380 - 12831410 on Build GRCh38
          Chromosome Location:
          Chr.1: 13315581 - 13322595 on Build GRCh38
          Chromosome Location:
          Chr.1: 13073678 - 13078038 on Build GRCh38
          Chromosome Location:
          Chr.1: 13148905 - 13153111 on Build GRCh38
          Chromosome Location:
          Chr.1: 13049808 - 13053659 on Build GRCh38
          Chromosome Location:
          Chr.1: 12879212 - 12886201 on Build GRCh38
          Chromosome Location:
          Chr.1: 13254198 - 13263435 on Build GRCh38
          Chromosome Location:
          Chr.1: 13175281 - 13179132 on Build GRCh38
          UniGene ID:
          Hs.567864
          Hs.551617
          Hs.551617
          -
          -
          -
          Hs.551617
          Hs.567864
          Hs.631873
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          -
          -
          -
          siRNA ID s54135

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PRAMEF6
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001010889.2 NP_001010889.1 4 1047
          GenBank mRNA BC101342.2 - 4 1047 40025149
          Gene Symbol: PRAMEF11
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001146344.2 NP_001139816.2 4 1038
          XM_011541479.2 XP_011539781.1 3 1224
          GenBank mRNA AL049680.1 - 4 1037
          BC144426.1 - 4 1037 9052947
          Gene Symbol: PRAMEF15
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001098376.2 NP_001091846.1 4 1061
          Gene Symbol: PRAMEF25
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001310134.1 NP_001297063.1 3 983
          Gene Symbol: PRAMEF26
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001306072.1 NP_001293001.1 3 983
          Gene Symbol: PRAMEF27
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001300891.1 NP_001287820.1 3 967
          Gene Symbol: PRAMEF4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001009611.4 NP_001009611.2 4 1038
          GenBank mRNA AL049681.1 - 4 1038
          Gene Symbol: PRAMEF5
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001013407.2 NP_001013425.2 4 1061
          GenBank mRNA BC101344.1 - 4 1047 40025152
          BC130274.1 - 4 1046 40146305
          BC130276.1 - 4 1046 40146307
          Gene Symbol: PRAMEF9
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001010890.2 NP_001010890.2 3 967
          GenBank mRNA AK124292.1 - 4 1061
          BC101343.2 - 4 1047 40025151
          BC137391.1 - 4 1047 9021389
          BC171794.1 - 4 1047 9054448

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of cell proliferation
          negative regulation of apoptotic process
          negative regulation of cell differentiation
          negative regulation of transcription, DNA-templated
          negative regulation of retinoic acid receptor signaling pathway
          retinoic acid receptor binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0