Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other FUT8 SIRNA Assays ›
          Gene Symbol:
          FUT8
          Gene Name:
          fucosyltransferase 8
          Gene Aliases:
          -
          Chromosome Location:
          Chr.14: 65410592 - 65744122 on Build GRCh38
          UniGene ID:
          Hs.597649
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s5424

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: FUT8
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_004480.4 NP_004471.4 10 2874
          NM_178155.2 NP_835368.1 11 3304
          NM_178156.2 NP_835369.1 10 3206
          NR_038167.1 9 2880
          NR_038170.1 11 2387
          XM_011536614.2 XP_011534916.1 6 1059
          XM_017021136.1 XP_016876625.1 12 2489
          XM_017021137.1 XP_016876626.1 12 2133
          XM_017021138.1 XP_016876627.1 11 2000
          XM_017021139.1 XP_016876628.1 11 2391
          XM_017021140.1 XP_016876629.1 10 4341
          GenBank mRNA AB049740.2 - 6 1446
          AF052088.1 - 5 943
          AJ514324.1 - 3 384
          AJ514325.1 - 3 384
          AJ536053.1 - 11 2772
          AJ536054.1 - 11 2772
          AJ536055.1 - 10 2674
          AJ536056.1 - 11 3302
          AJ539535.1 - 10 1902
          AJ539536.1 - 10 1902
          AK292413.1 - 11 2025
          AK294242.1 - 10 1592
          BC025385.2 - 11 1781
          BC093889.1 - 3 455
          BC101816.1 - 3 455
          BC142958.1 - 9 1607 8860361
          D89289.1 - 9 1719
          Y17976.1 - 11 2005
          Y17977.1 - 11 2005
          Y17978.1 - 11 2293
          Y17979.1 - 11 2293

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001002289, XM_008764825, XM_008764826, AJ781406, FN668938, and others.
          This siRNA also targets the following transcripts in Mouse: NM_001252614, NM_016893, NM_001252616, NM_001252615, EF591875, and others.

          Panther Classification:

          Molecular Function -

          glycosyltransferase

          Gene Ontology Categories:

          Function(s) Process(es)

          in utero embryonic development
          protein N-linked glycosylation
          N-glycan processing
          transforming growth factor beta receptor signaling pathway
          integrin-mediated signaling pathway
          respiratory gaseous exchange
          oligosaccharide biosynthetic process
          regulation of gene expression
          cell migration
          protein N-linked glycosylation via asparagine
          protein glycosylation in Golgi
          N-glycan fucosylation
          L-fucose catabolic process
          receptor metabolic process
          GDP-L-fucose metabolic process
          regulation of cellular response to oxidative stress
          glycoprotein 6-alpha-L-fucosyltransferase activity
          SH3 domain binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0