Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other LRRC37A2 SIRNA Assays ›
          Gene Symbol:
          LRRC37A2 LRRC37A LRRC37A3 LRRC37A4P
          Gene Name:
          leucine rich repeat containing 37 member A2
          leucine rich repeat containing 37A
          leucine rich repeat containing 37 member A3
          leucine rich repeat containing 37 member A4, pseudogene
          Gene Aliases:
          LRRC37
          LRRC37
          LRRC37, LRRC37A
          LRRC37, LRRC37A4
          Chromosome Location:
          Chr.17: 46512657 - 46556747 on Build GRCh38
          Chromosome Location:
          Chr.17: 46248044 - 46339367 on Build GRCh38
          Chromosome Location:
          Chr.17: 64854147 - 64966228 on Build GRCh38
          Chromosome Location:
          Chr.17: 45505883 - 45520523 on Build GRCh38
          UniGene ID:
          Hs.646568
          Hs.579108
          Hs.551962
          Hs.596756
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          siRNA ID s54594

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: LRRC37A2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001006607.2 NP_001006608.2 5 2845
          XM_011524841.2 XP_011523143.1 4 2826
          XM_011524842.2 XP_011523144.1 5 2898
          XM_011524843.2 XP_011523145.1 5 2898
          XM_011524849.1 XP_011523151.1 5 2898
          XM_011524850.2 XP_011523152.1 5 2898
          GenBank mRNA AK126994.1 - 5 1527
          AY386262.1 - 5 1266
          BC144427.1 - 5 508
          BC144428.1 - 5 508
          JQ790563.1 - 5 402
          JQ790564.1 - 5 402
          JQ790568.1 - 5 402
          JQ790570.1 - 5 402
          JQ790571.1 - 5 402
          JQ790576.1 - 5 402
          JQ790584.1 - 5 402
          JQ790598.1 - 4 330
          JQ790606.1 - 5 402
          JQ790610.1 - 5 402
          JQ790617.1 - 5 402
          JQ790626.1 - 5 402
          JQ790628.1 - 5 402
          JQ790700.1 - 5 402
          JQ790703.1 - 5 378
          JQ790735.1 - 5 404
          JQ790739.1 - 5 404
          JQ790740.1 - 5 404
          JQ790748.1 - 5 405
          JQ790750.1 - 5 404
          JQ790764.1 - 5 404
          Gene Symbol: LRRC37A
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_014834.4 NP_055649.4 5 2845
          XM_005257879.4 XP_005257936.1 5 3103
          XM_005257880.4 XP_005257937.1 5 3103
          XM_005257882.4 XP_005257939.1 6 3289
          XM_005257883.4 XP_005257940.1 5 3103
          XM_005257884.4 XP_005257941.1 5 3103
          XM_017025472.1 XP_016880961.1 7 2702
          XM_017025473.1 XP_016880962.1 5 349
          GenBank mRNA BC040501.1 - 5 2845 5295808
          CR749232.1 - 5 2846
          JQ790562.1 - 5 402
          JQ790566.1 - 5 402
          JQ790572.1 - 5 402
          JQ790577.1 - 5 402
          JQ790578.1 - 5 402
          JQ790579.1 - 5 402
          JQ790580.1 - 5 402
          JQ790581.1 - 5 402
          JQ790688.1 - 5 402
          JQ790690.1 - 5 402
          JQ790691.1 - 5 402
          JQ790692.1 - 5 402
          JQ790693.1 - 5 402
          JQ790694.1 - 5 402
          JQ790699.1 - 5 402
          JQ790726.1 - 5 404
          JQ790727.1 - 5 404
          JQ790729.1 - 5 404
          JQ790730.1 - 5 404
          JQ790738.1 - 5 404
          JQ790741.1 - 5 404
          JQ790743.1 - 5 404
          JQ790744.1 - 5 403
          JQ790749.1 - 5 404
          JQ790752.1 - 5 404
          JQ790755.1 - 5 404
          JQ790756.1 - 5 404
          JQ790757.1 - 5 404
          JQ790758.1 - 5 404
          JQ790763.1 - 5 405
          Gene Symbol: LRRC37A3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_199340.3 NP_955372.2 8 3508
          XM_011524776.1 XP_011523078.1 5 947
          XM_011524778.2 XP_011523080.2 6 472
          XM_017024593.1 XP_016880082.1 6 3337
          XM_017024594.1 XP_016880083.1 5 3265
          XM_017024595.1 XP_016880084.1 5 3265
          XM_017024599.1 XP_016880088.1 7 2709
          XM_017024600.1 XP_016880089.1 6 3337
          XM_017024601.1 XP_016880090.1 5 649
          XM_017024603.1 XP_016880092.1 5 947
          XM_017024604.1 XP_016880093.1 4 875
          XM_017024607.1 XP_016880096.1 5 252
          XM_017024608.1 XP_016880097.1 5 1269
          GenBank mRNA AB011135.2 - 7 3373
          AK315945.1 - 5 980
          BC043145.1 - 5 2850 5296147
          BI560889.1 - 5 289
          JQ790603.1 - 5 402
          JQ790607.1 - 5 402
          JQ790608.1 - 5 402
          JQ790612.1 - 5 402
          JQ790613.1 - 5 402
          JQ790704.1 - 5 402
          JQ790705.1 - 5 402
          JQ790706.1 - 5 402
          JQ790713.1 - 4 330
          JQ790731.1 - 5 404
          JQ790734.1 - 5 412
          JQ790736.1 - 4 275
          JQ790737.1 - 5 404
          JQ790747.1 - 5 404
          JQ790751.1 - 5 404
          JQ790753.1 - 5 404
          JQ790759.1 - 5 404
          JQ790760.1 - 5 402
          JQ790761.1 - 5 404
          JQ790762.1 - 5 405
          JQ790765.1 - 5 404
          JQ790766.1 - 5 404
          Gene Symbol: LRRC37A4P
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA JQ790585.1 - 5 402
          JQ790586.1 - 5 402
          JQ790630.1 - 5 402
          JQ790698.1 - 5 402

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0