Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other GAK SIRNA Assays ›
          Gene Symbol:
          GAK
          Gene Name:
          cyclin G associated kinase
          Gene Aliases:
          DNAJ26, DNAJC26
          Chromosome Location:
          Chr.4: 849275 - 932390 on Build GRCh38
          UniGene ID:
          Hs.369607
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s5527

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: GAK
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001318134.1 NP_001305063.1 8, 9 1168
          NM_005255.3 NP_005246.2 11, 12 1405
          XM_005272268.2 XP_005272325.1 11, 12 1345
          XM_005272270.2 XP_005272327.1 11, 12 1345
          XM_011513425.2 XP_011511727.1 11, 12 1345
          XM_011513426.2 XP_011511728.1 11, 12 1345
          XM_011513427.2 XP_011511729.1 10, 11 1255
          XM_011513428.2 XP_011511730.1 11, 12 1345
          XM_011513429.2 XP_011511731.1 9, 10 1667
          XM_011513430.1 XP_011511732.1 8, 9 1092
          XM_011513431.2 XP_011511733.1 11, 12 1345
          XM_011513432.2 XP_011511734.1 11, 12 1316
          XM_011513434.2 XP_011511736.1 9, 10 1157
          XM_017007991.1 XP_016863480.1 10, 11 1255
          XM_017007992.1 XP_016863481.1 7, 8 1002
          XM_017007993.1 XP_016863482.1 9, 10 1094
          XM_017007994.1 XP_016863483.1 11, 12 1345
          XM_017007995.1 XP_016863484.1 10, 11 1164
          GenBank mRNA AK299554.1 - 8, 9 1040
          AK309413.1 - 8, 9 1024
          BC085005.1 - 8, 9 1151 6598984
          D88435.1 - 11, 12 1202

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          membrane traffic protein

          Gene Ontology Categories:

          Function(s) Process(es)

          protein phosphorylation
          receptor-mediated endocytosis
          endoplasmic reticulum organization
          Golgi organization
          cell cycle
          negative regulation of neuron projection development
          clathrin coat disassembly
          clathrin-mediated endocytosis
          establishment of protein localization to Golgi
          establishment of protein localization to plasma membrane
          Golgi to lysosome transport
          protein serine/threonine kinase activity
          protein binding
          ATP binding
          cyclin binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-mcfj8:80/100.66.77.12:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0