Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other LRRC37A SIRNA Assays ›
          Gene Symbol:
          LRRC37A LRRC37A2 LRRC37A3
          Gene Name:
          leucine rich repeat containing 37A
          leucine rich repeat containing 37 member A2
          leucine rich repeat containing 37 member A3
          Gene Aliases:
          LRRC37
          LRRC37
          LRRC37, LRRC37A
          Chromosome Location:
          Chr.17: 46248044 - 46339367 on Build GRCh38
          Chromosome Location:
          Chr.17: 46512657 - 46556747 on Build GRCh38
          Chromosome Location:
          Chr.17: 64854147 - 64966228 on Build GRCh38
          UniGene ID:
          Hs.579108
          Hs.646568
          Hs.551962
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          siRNA ID s55661

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 392.00
          Your Price
          Online offer: 380.65
          380.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: LRRC37A
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_014834.4 NP_055649.4 1 674
          XM_005257879.4 XP_005257936.1 1 932
          XM_005257880.4 XP_005257937.1 1 932
          XM_005257882.4 XP_005257939.1 2 1118
          XM_005257883.4 XP_005257940.1 1 932
          XM_005257884.4 XP_005257941.1 1 932
          XM_006722210.3 XP_006722273.1 1 932
          XM_006722211.3 XP_006722274.1 1 932
          XM_011525539.2 XP_011523841.1 1 932
          XM_017025472.1 XP_016880961.1 3 531
          GenBank mRNA BC040501.1 - 1 674 5295808
          CR749232.1 - 1 675
          Gene Symbol: LRRC37A2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001006607.2 NP_001006608.2 1 674
          XM_011524841.2 XP_011523143.1 1 727
          XM_011524842.2 XP_011523144.1 1 727
          XM_011524843.2 XP_011523145.1 1 727
          XM_011524844.2 XP_011523146.1 1 727
          XM_011524846.2 XP_011523148.1 1 727
          XM_011524848.2 XP_011523150.1 1 727
          XM_011524849.1 XP_011523151.1 1 727
          XM_011524850.2 XP_011523152.1 1 727
          Gene Symbol: LRRC37A3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_199340.3 NP_955372.2 4 1337
          XM_017024593.1 XP_016880082.1 2 1166
          XM_017024594.1 XP_016880083.1 2 1166
          XM_017024595.1 XP_016880084.1 2 1166
          XM_017024596.1 XP_016880085.1 2 1166
          XM_017024597.1 XP_016880086.1 2 1166
          XM_017024598.1 XP_016880087.1 2 1166
          XM_017024599.1 XP_016880088.1 3 538
          XM_017024600.1 XP_016880089.1 2 1166
          GenBank mRNA AB011135.2 - 3 1202
          BC043145.1 - 1 679 5296147

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0