Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PABPC1L2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042506.1 | NP_001035971.1 | 1 | 622 | ||
GenBank mRNA | BC041956.1 | - | 1 | 622 | ||
BC136492.1 | - | 1 | 525 | 9020481 |
Gene Symbol: PABPC1L2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012977.2 | NP_001012995.1 | 1 | 618 |