Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HERC2 SIRNA Assays ›
          Gene Symbol:
          HERC2 HERC2P2 HERC2P3 HERC2P9 LOC102723753
          Gene Name:
          HECT and RLD domain containing E3 ubiquitin protein ligase 2
          hect domain and RLD 2 pseudogene 2
          hect domain and RLD 2 pseudogene 3
          hect domain and RLD 2 pseudogene 9
          HECT and RLD domain containing E3 ubiquitin protein ligase 2 pseudogene
          Gene Aliases:
          D15F37S1, MRT38, SHEP1, jdf2, p528
          D15F37S3, MN7
          D15F37S4
          -
          -
          Chromosome Location:
          Chr.15: 28111037 - 28322173 on Build GRCh38
          Chromosome Location:
          Chr.15: 22494837 - 22590831 on Build GRCh38
          Chromosome Location:
          Chr.15: 20408396 - 20506180 on Build GRCh38
          Chromosome Location:
          Chr.15: 28654442 - 28685264 on Build GRCh38
          Chromosome Location:
          Chr.16|NT_187383: 1001630 - 1037618 on Build GRCh38
          UniGene ID:
          Hs.434890
          Hs.458334
          Hs.608918
          Hs.531509
          -
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          siRNA ID s57297

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: HERC2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_004667.5 NP_004658.3 31 4899
          XM_005268276.4 XP_005268333.1 30 5029
          XM_006720726.3 XP_006720789.1 31 4905
          XM_006720727.3 XP_006720790.1 29 4662
          XM_011522131.2 XP_011520433.1 29 4686
          XM_011522133.2 XP_011520435.1 12 1665
          XM_017022695.1 XP_016878184.1 30 4846
          XM_017022696.1 XP_016878185.1 30 4789
          GenBank mRNA AF071172.3 - 31 4851
          Gene Symbol: HERC2P2
          Interrogated Sequence Translated Protein RNADB ID Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_002824.3
          12 1726
          GenBank mRNA AB002391.2 -
          10 1471
          AF041080.1 - LIT2190
          HERC2P2
          12 1730
          AF041081.1 - LIT2191
          HERC2P3
          10 1406
          AK307870.1 -
          7 1108
          AK309255.1 -
          6 1384
          Gene Symbol: HERC2P3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_036432.1 11 1565
          GenBank mRNA AK294470.1 - 6 986
          AK294795.1 - 7 1111
          BC033888.1 - 11 1570
          BC094757.1 - 11 1565
          CN398998.1 - 2 278
          Gene Symbol: HERC2P9
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA BM464322.1 - 5 715
          Gene Symbol: LOC102723753
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_135178.1 15 2328

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          guanyl-nucleotide exchange factor

          Gene Ontology Categories:

          Function(s) Process(es)

          double-strand break repair via nonhomologous end joining
          intracellular protein transport
          cellular response to DNA damage stimulus
          spermatogenesis
          protein ubiquitination
          protein sumoylation
          protein ubiquitination involved in ubiquitin-dependent protein catabolic process
          positive regulation of GTPase activity
          ubiquitin-protein transferase activity
          guanyl-nucleotide exchange factor activity
          protein binding
          zinc ion binding
          ligase activity
          ubiquitin protein ligase binding
          SUMO binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0