Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: FAM72D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207418.2 | NP_997301.2 | 4 | 1245 | ||
XM_011509962.2 | XP_011508264.1 | 5 | 1280 | |||
XM_017002227.1 | XP_016857716.1 | 6 | 1665 | |||
GenBank mRNA | AB096683.1 | - | 4 | 1245 | ||
AK307197.1 | - | 3 | 1096 |
Gene Symbol: FAM72A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001123168.2 | NP_001116640.1 | 4 | 1247 | ||
NM_001317901.1 | NP_001304830.1 | 4 | 467 | |||
NR_134239.1 | 5 | 527 | ||||
XM_006711526.3 | XP_006711589.1 | 5 | 1221 | |||
XM_011509965.2 | XP_011508267.1 | 5 | 1130 | |||
XM_011509967.1 | XP_011508269.1 | 4 | 1142 | |||
XM_017002233.1 | XP_016857722.1 | 5 | 1127 | |||
GenBank mRNA | BC035696.1 | - | 4 | 814 | 5541342 | |
BC137545.1 | - | 4 | 778 | 9021545 | ||
BC146978.1 | - | 4 | 1246 | 9056906 | ||
BC146992.1 | - | 4 | 1245 | 9056920 | ||
BI916384.1 | - | 4 | 468 |
Gene Symbol: FAM72B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100910.1 | NP_001094380.1 | 4 | 1226 | ||
NM_001320149.1 | NP_001307078.1 | 4 | 699 | |||
XM_011542018.1 | XP_011540320.1 | 5 | 1634 | |||
GenBank mRNA | BC046199.1 | - | 4 | 699 | 6064384 | |
BC137546.1 | - | 4 | 779 | 9021546 |
Gene Symbol: FAM72C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287385.1 | NP_001274314.1 | 4 | 1262 | ||
XM_011509723.2 | XP_011508025.1 | 5 | 1315 | |||
XM_017001682.1 | XP_016857171.1 | 5 | 1512 |