Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SRSF10 SIRNA Assays ›
          Gene Symbol:
          SRSF10
          Gene Name:
          serine and arginine rich splicing factor 10
          Gene Aliases:
          FUSIP1, FUSIP2, NSSR, PPP1R149, SFRS13, SFRS13A, SRp38, SRrp40, TASR, TASR1, TASR2
          Chromosome Location:
          Chr.1: 23964347 - 23980927 on Build GRCh38
          UniGene ID:
          Hs.3530
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s57796

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SRSF10
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001191005.2 NP_001177934.1 3 419
          NM_001191006.2 NP_001177935.1 3 419
          NM_001191007.2 NP_001177936.1 3 419
          NM_001191009.2 NP_001177938.1 3 419
          NM_001300936.1 NP_001287865.1 3 419
          NM_001300937.1 NP_001287866.1 3 419
          NM_006625.5 NP_006616.1 3 419
          NM_054016.3 NP_473357.1 3 419
          NR_034035.2 4 816
          XM_017000100.1 XP_016855589.1 3 4139
          XM_017000101.1 XP_016855590.1 4 3682
          XM_017000102.1 XP_016855591.1 4 1309
          GenBank mRNA AF047448.1 - 3 240
          AF067730.1 - 3 240
          AF419331.1 - 3 287
          AF449427.1 - 3 211
          AK001286.1 - 3 283
          AK001656.1 - 3 283
          AK055311.1 - 1 1221
          AK093226.1 - 4 1280
          AK125834.1 - 1 2616
          AK222994.1 - 3 310
          AK296175.1 - 3 312
          AY007101.1 - 3 252
          AY150180.1 - 3 225
          AY150181.1 - 3 225
          BC001107.1 - 3 312 3504780
          BC005039.1 - 3 275 3531441
          BC010074.2 - 3 264 4302658
          BC071575.1 - 3 3540
          BX648939.1 - 1 2472
          KU178536.1 - 3 211

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001025738, XM_006239220, XM_006239222, XM_006239223, XM_006239221, and others.
          This siRNA also targets the following transcripts in Mouse: NM_001284195, NM_001284196, NM_001080387, NM_010178, BC006670, and others.

          Panther Classification:

          Molecular Function -

          RNA splicing factor

          Gene Ontology Categories:

          Function(s) Process(es)

          spliceosomal tri-snRNP complex assembly
          RNA splicing, via transesterification reactions
          mRNA splicing, via spliceosome
          regulation of transcription, DNA-templated
          mRNA splice site selection
          mRNA export from nucleus
          cytosolic transport
          regulation of mRNA splicing, via spliceosome
          negative regulation of mRNA splicing, via spliceosome
          nucleotide binding
          RNA binding
          protein binding
          poly(A) RNA binding
          RS domain binding
          unfolded protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0