Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PARG SIRNA Assays ›
          Gene Symbol:
          PARG PARGP1
          Gene Name:
          poly(ADP-ribose) glycohydrolase
          poly(ADP-ribose) glycohydrolase pseudogene 1
          Gene Aliases:
          PARG99
          -
          Chromosome Location:
          Chr.10: 49818275 - 49942027 on Build GRCh38
          Chromosome Location:
          Chr.10: 45854835 - 45972428 on Build GRCh38
          UniGene ID:
          Hs.535298
          Hs.499594
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID s57869

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PARG
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001303486.1 NP_001290415.1 6, 7 1533
          NM_001303487.1 NP_001290416.1 5, 6 1896
          NM_001303489.1 NP_001290418.1 3, 4 725
          NM_001324381.1 NP_001311310.1 6, 7 1537
          NM_003631.3 NP_003622.2 6, 7 1963
          NR_130168.1 6, 7 1963
          NR_130169.1 4, 5 909
          NR_136752.1 5, 6 1896
          NR_136753.1 5, 6 976
          NR_136754.1 5, 6 1896
          NR_136755.1 6, 7 1963
          NR_136756.1 6, 7 1963
          XM_011540305.1 XP_011538607.1 5, 6 1806
          XM_011540308.2 XP_011538610.1 6, 7 1929
          GenBank mRNA AF005043.1 - 7, 8 1827
          AK295786.1 - 5, 6 1855
          AK301845.1 - 4, 5 855
          AK302560.1 - 6, 7 1504
          AY258587.1 - 6, 7 1922
          AY575848.1 - 6, 7 1517
          AY575849.1 - 5, 6 1428
          BC050560.1 - 6, 7 1889 6064831
          BC052966.1 - 6, 7 1877 6042331
          DQ867088.1 - 6, 7 1741
          EF382674.1 - 3, 4 466
          JQ890226.1 - 3, 4 394
          Gene Symbol: PARGP1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_029388.2 3, 4 479
          GenBank mRNA AK311609.1 - 3, 4 451

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          glycosidase

          Gene Ontology Categories:

          Function(s) Process(es)

          carbohydrate metabolic process
          ATP generation from poly-ADP-D-ribose
          poly(ADP-ribose) glycohydrolase activity
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-w8bxd:80/100.66.79.51:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0