Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CCR5 SIRNA Assays ›
          Gene Symbol:
          CCR5
          Gene Name:
          C-C motif chemokine receptor 5 (gene/pseudogene)
          Gene Aliases:
          CC-CKR-5, CCCKR5, CCR-5, CD195, CKR-5, CKR5, CMKBR5, IDDM22
          Chromosome Location:
          Chr.3: 46370142 - 46376206 on Build GRCh38
          UniGene ID:
          Hs.450802
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s57926

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CCR5
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000579.3 NP_000570.1 3 682
          NM_001100168.1 NP_001093638.1 3 447
          GenBank mRNA AF011500.1 - 1 329
          AF011501.1 - 1 329
          AF011502.1 - 1 329
          AF011503.1 - 1 329
          AF011504.1 - 1 328
          AF011505.1 - 1 329
          AF011506.1 - 1 329
          AF011507.1 - 1 329
          AF011508.1 - 1 329
          AF011509.1 - 1 329
          AF011510.1 - 1 329
          AF011511.1 - 1 329
          AF011512.1 - 1 329
          AF011513.1 - 1 329
          AF011514.1 - 1 329
          AF011515.1 - 1 329
          AF011516.1 - 1 329
          AF011517.1 - 1 329
          AF011518.1 - 1 329
          AF011519.1 - 1 329
          AF011520.1 - 1 329
          AF011521.1 - 1 329
          AF011522.1 - 1 329
          AF011523.1 - 1 329
          AF011524.1 - 1 329
          AF011525.1 - 1 329
          AF011526.1 - 1 329
          AF011527.1 - 1 329
          AF011528.1 - 1 329
          AF011529.1 - 1 329
          AF011530.1 - 1 329
          AF011531.1 - 1 329
          AF011532.1 - 1 329
          AF011533.1 - 1 329
          AF011534.1 - 1 329
          AF011535.1 - 1 329
          AF011536.1 - 1 329
          AF011537.1 - 1 329
          AK292132.1 - 2 379
          AK314882.1 - 2 471
          BC038398.1 - 2 368 5186388
          DA818906.1 - 2 471
          U54994.1 - 2 379
          U57840.1 - 2 351

          Back To Top

          More Information


          Set Membership:

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          dendritic cell chemotaxis
          calcium ion transport
          chemotaxis
          inflammatory response
          immune response
          cellular defense response
          cell surface receptor signaling pathway
          G-protein coupled receptor signaling pathway
          positive regulation of cytosolic calcium ion concentration
          cell-cell signaling
          release of sequestered calcium ion into cytosol by sarcoplasmic reticulum
          fusion of virus membrane with host plasma membrane
          calcium-mediated signaling
          signaling
          entry into host cell
          chemokine-mediated signaling pathway
          response to cholesterol
          cellular response to lipopolysaccharide
          virus receptor activity
          actin binding
          phosphatidylinositol phospholipase C activity
          chemokine receptor activity
          protein binding
          coreceptor activity
          C-C chemokine receptor activity
          C-C chemokine binding
          chemokine (C-C motif) ligand 5 binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-nmj5c:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0