Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NDST1 SIRNA Assays ›
Gene Symbol:
NDST1
Gene Name:
N-deacetylase and N-sulfotransferase 1
Gene Aliases:
HSST, MRT46, NST1
Chromosome Location:
Chr.5: 150485783 - 150558211 on Build GRCh38
UniGene ID:
Hs.222055
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s60
Validated
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
Availability
Inventoried
Catalog #
4390824
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NDST1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001301063.1
NP_001287992.1
7, 8
2147
NM_001543.4
NP_001534.1
7, 8
2060
XM_005268433.1
XP_005268490.1
7, 8
2032
XM_005268434.2
XP_005268491.1
7, 8
2641
XM_005268435.4
XP_005268492.1
7, 8
2166
XM_005268436.3
XP_005268493.1
7, 8
3806
XM_005268437.4
XP_005268494.1
7, 8
2045
XM_005268438.1
XP_005268495.1
7, 8
2032
XM_005268439.1
XP_005268496.1
7, 8
2032
XM_005268442.3
XP_005268499.1
5, 6
1015
XM_006714782.1
XP_006714845.1
7, 8
2067
XM_006714783.1
XP_006714846.1
7, 8
2032
XM_011537638.2
XP_011535940.1
6, 7
1122
XM_017009427.1
XP_016864916.1
7, 8
5023
XM_017009428.1
XP_016864917.1
7, 8
2165
XM_017009429.1
XP_016864918.1
7, 8
2045
XM_017009430.1
XP_016864919.1
7, 8
2067
XM_017009431.1
XP_016864920.1
7, 8
2637
XM_017009432.1
XP_016864921.1
6, 7
3818
XM_017009433.1
XP_016864922.1
6, 7
3816
GenBank mRNA
AB209107.1
-
7, 8
1834
AK292448.1
-
7, 8
2060
AK293746.1
-
7, 8
2147
U17970.1
-
6, 7
1786
U18918.1
-
6, 7
1774
U36600.1
-
7, 8
1981
Back To Top
More Information
Matching Taqman Gene Expression Assay:
Hs00155454_m1
Panther Classification:
Molecular Function -
transferase
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
polysaccharide biosynthetic process
cardiac septum development
glycosaminoglycan biosynthetic process
protein sulfation
inflammatory response
smoothened signaling pathway
respiratory gaseous exchange
fibroblast growth factor receptor signaling pathway
heparan sulfate proteoglycan biosynthetic process
heparin biosynthetic process
forebrain development
midbrain development
aorta development
embryonic neurocranium morphogenesis
embryonic viscerocranium morphogenesis
coronary vasculature development
protein binding
[heparan sulfate]-glucosamine N-sulfotransferase activity
N-acetylglucosamine deacetylase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-hqkpn:80/100.66.73.1:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0