Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NDST1 SIRNA Assays ›
          Gene Symbol:
          NDST1
          Gene Name:
          N-deacetylase and N-sulfotransferase 1
          Gene Aliases:
          HSST, MRT46, NST1
          Chromosome Location:
          Chr.5: 150485783 - 150558211 on Build GRCh38
          UniGene ID:
          Hs.222055
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s61

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 392.00
          Your Price
          Online offer: 380.65
          380.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NDST1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001301063.1 NP_001287992.1 11 2608
          NM_001543.4 NP_001534.1 11 2521
          XM_005268433.1 XP_005268490.1 11 2493
          XM_005268434.2 XP_005268491.1 11 3102
          XM_005268435.4 XP_005268492.1 11 2627
          XM_005268436.3 XP_005268493.1 11 4267
          XM_005268437.4 XP_005268494.1 11 2506
          XM_005268438.1 XP_005268495.1 11 2493
          XM_005268439.1 XP_005268496.1 11 2493
          XM_005268442.3 XP_005268499.1 9 1476
          XM_006714782.1 XP_006714845.1 11 2528
          XM_006714783.1 XP_006714846.1 11 2493
          XM_011537638.2 XP_011535940.1 10 1583
          XM_017009427.1 XP_016864916.1 11 5484
          XM_017009428.1 XP_016864917.1 11 2626
          XM_017009429.1 XP_016864918.1 11 2506
          XM_017009430.1 XP_016864919.1 11 2528
          XM_017009431.1 XP_016864920.1 11 3098
          XM_017009432.1 XP_016864921.1 10 4279
          XM_017009433.1 XP_016864922.1 10 4277
          GenBank mRNA AB209107.1 - 11 2295
          AK292448.1 - 11 2521
          AK293746.1 - 11 2608
          U17970.1 - 10 2247
          U18918.1 - 10 2235
          U36600.1 - 11 2442

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          transferase

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          polysaccharide biosynthetic process
          cardiac septum development
          glycosaminoglycan biosynthetic process
          protein sulfation
          inflammatory response
          smoothened signaling pathway
          respiratory gaseous exchange
          fibroblast growth factor receptor signaling pathway
          heparan sulfate proteoglycan biosynthetic process
          heparin biosynthetic process
          forebrain development
          midbrain development
          aorta development
          embryonic neurocranium morphogenesis
          embryonic viscerocranium morphogenesis
          coronary vasculature development
          protein binding
          [heparan sulfate]-glucosamine N-sulfotransferase activity
          N-acetylglucosamine deacetylase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-2rf75:80/100.66.79.83:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0