Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other OBSCN SIRNA Assays ›
Gene Symbol:
OBSCN
Gene Name:
obscurin, cytoskeletal calmodulin and titin-interacting RhoGEF
Gene Aliases:
ARHGEF30, UNC89
Chromosome Location:
Chr.1: 228208013 - 228381431 on Build GRCh38
UniGene ID:
Hs.650039
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s61236
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: OBSCN
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001098623.2
NP_001092093.2
89
19990
NM_001271223.2
NP_001258152.2
100
22861
XM_005273287.4
XP_005273344.1
101
23247
XM_005273291.4
XP_005273348.1
101
23211
XM_005273298.4
XP_005273355.1
100
22938
XM_005273307.4
XP_005273364.1
96
22125
XM_006711822.3
XP_006711885.1
99
22671
XM_006711823.3
XP_006711886.1
99
22665
XM_011544297.2
XP_011542599.1
93
21081
XM_011544299.2
XP_011542601.1
92
20805
XM_017002443.1
XP_016857932.1
101
23292
XM_017002444.1
XP_016857933.1
101
23255
XM_017002445.1
XP_016857934.1
101
23289
XM_017002446.1
XP_016857935.1
101
23289
XM_017002447.1
XP_016857936.1
101
23289
XM_017002448.1
XP_016857937.1
101
23142
XM_017002449.1
XP_016857938.1
100
23029
XM_017002450.1
XP_016857939.1
100
23029
XM_017002451.1
XP_016857940.1
100
23029
XM_017002452.1
XP_016857941.1
100
23029
XM_017002453.1
XP_016857942.1
100
23016
XM_017002454.1
XP_016857943.1
100
23016
XM_017002455.1
XP_016857944.1
100
23016
XM_017002456.1
XP_016857945.1
100
23016
XM_017002457.1
XP_016857946.1
100
23016
XM_017002458.1
XP_016857947.1
99
22765
XM_017002459.1
XP_016857948.1
99
22765
XM_017002460.1
XP_016857949.1
99
22765
XM_017002461.1
XP_016857950.1
99
22752
XM_017002462.1
XP_016857951.1
99
22740
XM_017002463.1
XP_016857952.1
99
22719
XM_017002464.1
XP_016857953.1
99
22668
XM_017002465.1
XP_016857954.1
98
22501
XM_017002466.1
XP_016857955.1
98
22501
XM_017002467.1
XP_016857956.1
98
22501
XM_017002468.1
XP_016857957.1
97
22237
XM_017002469.1
XP_016857958.1
95
21709
XM_017002470.1
XP_016857959.1
90
20390
GenBank mRNA
AB046859.2
-
29
3765
AM231061.1
-
15
1892
DQ400343.1
-
15
1892
Back To Top
More Information
Panther Classification:
Molecular Function -
immunoglobulin
Gene Ontology Categories:
Function(s)
Process(es)
protein phosphorylation
multicellular organism development
regulation of Rho protein signal transduction
protein localization to M-band
positive regulation of apoptotic process
positive regulation of GTPase activity
sarcomere organization
regulation of small GTPase mediated signal transduction
mitophagy in response to mitochondrial depolarization
protein serine/threonine kinase activity
guanyl-nucleotide exchange factor activity
Rho guanyl-nucleotide exchange factor activity
protein binding
calmodulin binding
ATP binding
structural constituent of muscle
ankyrin binding
titin binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-qm6d9:80/100.66.79.83:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline