Search
Search

Abl1| Gene Symbol: Abl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001112703.2 | NP_001106174.1 | 11 | 4539 | ||
| NM_001283045.1 | NP_001269974.1 | 11 | 3416 | |||
| NM_001283046.1 | NP_001269975.1 | 12 | 4744 | |||
| NM_001283047.1 | NP_001269976.1 | 12 | 4683 | |||
| NM_009594.4 | NP_033724.2 | 11 | 3310 | |||
| GenBank mRNA | AK090095.1 | - | 11 | 3268 | ||
| BC013801.1 | - | 1 | 939 | |||
| BC035521.1 | - | 1 | 51 | |||
| BC059260.1 | - | 11 | 3337 | 6408990 | ||
| BC103770.1 | - | 11 | 3763 | 30848553 | ||
| BM949741.1 | - | 1 | 363 | |||
| CA752052.1 | - | 1 | 273 | |||
| J02995.1 | - | 11 | 3263 | |||
| L10656.1 | - | 9 | 2864 | |||