Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ADORA2A SIRNA Assays ›
          Gene Symbol:
          Adora2a
          Gene Name:
          adenosine A2a receptor
          Gene Aliases:
          A2AAR, A2aR, AA2AR
          Chromosome Location:
          Chr.10: 75316860 - 75334792 on Build GRCm38
          UniGene ID:
          Mm.333734
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID s62045

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price 5,354.00
          Your Price
          Online offer:
          5354.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Adora2a
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_009630.3 NP_033760.2 3 1121
          GenBank mRNA AK037787.1 - 2 343
          BC110692.1 - 2 621 30242398
          Y13346.1 - 2 606

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_053294, BC081727, S47609, M91214, L08102, and others.

          Set Membership:

          G-protein coupled receptor

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          synaptic transmission, dopaminergic
          adenosine receptor signaling pathway
          response to amphetamine
          regulation of transcription, DNA-templated
          negative regulation of protein kinase activity
          signal transduction
          G-protein coupled receptor signaling pathway
          adenylate cyclase-activating G-protein coupled receptor signaling pathway
          protein kinase C-activating G-protein coupled receptor signaling pathway
          synaptic transmission, cholinergic
          locomotory behavior
          negative regulation of cell proliferation
          positive regulation of adenylate cyclase activity involved in G-protein coupled receptor signaling pathway
          positive regulation of glutamate secretion
          positive regulation of acetylcholine secretion, neurotransmission
          regulation of norepinephrine secretion
          positive regulation of cAMP biosynthetic process
          response to caffeine
          positive regulation of synaptic transmission, GABAergic
          synaptic transmission, glutamatergic
          positive regulation of urine volume
          positive regulation of renal sodium excretion
          negative regulation of locomotion
          vasodilation
          eating behavior
          negative regulation of vascular permeability
          negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
          response to alkaloid
          negative regulation of neuron apoptotic process
          positive regulation of circadian sleep/wake cycle, sleep
          negative regulation of alpha-beta T cell activation
          astrocyte activation
          regulation of synaptic plasticity
          neuron projection morphogenesis
          positive regulation of protein secretion
          negative regulation of inflammatory response
          regulation of mitochondrial membrane potential
          membrane depolarization
          regulation of calcium ion transport
          positive regulation of synaptic transmission, glutamatergic
          excitatory postsynaptic potential
          inhibitory postsynaptic potential
          prepulse inhibition
          positive regulation of apoptotic signaling pathway
          G-protein coupled adenosine receptor activity
          signal transducer activity
          G-protein coupled receptor activity
          enzyme binding
          type 5 metabotropic glutamate receptor binding
          identical protein binding
          protein heterodimerization activity
          alpha-actinin binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0